Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33768672-33768939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO3
Entrez Gene ID
26273
Gene Name
F-box protein 3
Gene Aliases
FBA, FBX3
UniGene
Hs.406787
Gene Chromosome Location
Chr.11: 33740937-33774531 on Build GRCh38
Transcripts
NM_012175.3 NM_033406.2 XM_011519980.1 XM_011519981.2 XM_011519982.1 XM_017017533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3446887 c.309C>T p.L103L Chr.11: 33768900-33768900 on Build GRCh38
3739031 c.275A>G p.H92R Chr.11: 33768934-33768934 on Build GRCh38
3446886 c.335C>T p.P112L Chr.11: 33768874-33768874 on Build GRCh38
6260422 c.325C>A p.P109T Chr.11: 33768884-33768884 on Build GRCh38
4032488 c.277T>C p.Y93H Chr.11: 33768932-33768932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746147943 Chr.11: 33768869-33768869 on Build GRCh38
rs747495018 Chr.11: 33768890-33768890 on Build GRCh38
rs779204780 Chr.11: 33768864-33768864 on Build GRCh38
rs187370840 Chr.11: 33768730-33768730 on Build GRCh38
rs769264685 Chr.11: 33768908-33768908 on Build GRCh38
rs566300401 Chr.11: 33768735-33768735 on Build GRCh38
rs927410629 Chr.11: 33768738-33768738 on Build GRCh38
rs762812423 Chr.11: 33768762-33768762 on Build GRCh38
rs969993621 Chr.11: 33768768-33768768 on Build GRCh38
rs766267700 Chr.11: 33768778-33768778 on Build GRCh38
rs145480518 Chr.11: 33768857-33768857 on Build GRCh38
rs928527495 Chr.11: 33768782-33768782 on Build GRCh38
rs776328124 Chr.11: 33768753-33768753 on Build GRCh38
rs190001510 Chr.11: 33768822-33768822 on Build GRCh38
rs891379530 Chr.11: 33768823-33768823 on Build GRCh38
rs768302775 Chr.11: 33768752-33768752 on Build GRCh38
rs754960224 Chr.11: 33768796-33768796 on Build GRCh38
rs1915894 Chr.11: 33768722-33768722 on Build GRCh38
rs761369938 Chr.11: 33768757-33768757 on Build GRCh38
rs978421623 Chr.11: 33768867-33768867 on Build GRCh38
rs139067802 Chr.11: 33768734-33768737 on Build GRCh38
rs769324553 Chr.11: 33768891-33768891 on Build GRCh38
rs753560846 Chr.11: 33768783-33768783 on Build GRCh38
rs373693259 Chr.11: 33768721-33768721 on Build GRCh38
rs78742766 Chr.11: 33768709-33768709 on Build GRCh38
rs754014558 Chr.11: 33768829-33768829 on Build GRCh38
rs535750253 Chr.11: 33768774-33768774 on Build GRCh38
rs772595860 Chr.11: 33768871-33768871 on Build GRCh38
rs897290856 Chr.11: 33768783-33768783 on Build GRCh38
rs760221560 Chr.11: 33768750-33768750 on Build GRCh38
rs930226303 Chr.11: 33768787-33768787 on Build GRCh38
rs775993823 Chr.11: 33768894-33768894 on Build GRCh38
rs780657526 Chr.11: 33768884-33768884 on Build GRCh38
rs750128654 Chr.11: 33768761-33768761 on Build GRCh38
rs751524437 Chr.11: 33768792-33768792 on Build GRCh38
rs555339470 Chr.11: 33768775-33768775 on Build GRCh38
rs372913089 Chr.11: 33768758-33768758 on Build GRCh38
rs754247969 Chr.11: 33768711-33768711 on Build GRCh38
rs1016102184 Chr.11: 33768701-33768701 on Build GRCh38
rs11547296 Chr.11: 33768883-33768883 on Build GRCh38

View Full Product Details