Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49804087-49804360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM212A
Entrez Gene ID
389119
Gene Name
family with sequence similarity 212 member A
Gene Aliases
C3orf54, INKA1
UniGene
Hs.86674
Gene Chromosome Location
Chr.3: 49803130-49805030 on Build GRCh38
Transcripts
NM_203370.1 XM_017006372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7271948 c.232G>T p.E78* Chr.3: 49804355-49804355 on Build GRCh38
6659440 c.74C>T p.T25I Chr.3: 49804197-49804197 on Build GRCh38
7295230 c.224T>C p.V75A Chr.3: 49804347-49804347 on Build GRCh38
4682912 c.205_207delGAG p.E73delE Chr.3: 49804328-49804330 on Build GRCh38
6344829 c.175G>T p.D59Y Chr.3: 49804298-49804298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755837284 Chr.3: 49804191-49804191 on Build GRCh38
rs765680786 Chr.3: 49804139-49804139 on Build GRCh38
rs748831158 Chr.3: 49804300-49804300 on Build GRCh38
rs750575767 Chr.3: 49804154-49804154 on Build GRCh38
rs748167846 Chr.3: 49804184-49804184 on Build GRCh38
rs971658130 Chr.3: 49804128-49804128 on Build GRCh38
rs780851444 Chr.3: 49804284-49804284 on Build GRCh38
rs915049748 Chr.3: 49804200-49804200 on Build GRCh38
rs141243520 Chr.3: 49804324-49804324 on Build GRCh38
rs771241593 Chr.3: 49804314-49804315 on Build GRCh38
rs774567373 Chr.3: 49804328-49804330 on Build GRCh38
rs755029617 Chr.3: 49804282-49804282 on Build GRCh38
rs762032249 Chr.3: 49804138-49804138 on Build GRCh38
rs745329684 Chr.3: 49804226-49804226 on Build GRCh38
rs766605315 Chr.3: 49804166-49804166 on Build GRCh38
rs766552277 Chr.3: 49804264-49804264 on Build GRCh38
rs983318205 Chr.3: 49804152-49804152 on Build GRCh38
rs150234202 Chr.3: 49804334-49804334 on Build GRCh38
rs753618763 Chr.3: 49804317-49804317 on Build GRCh38
rs145857836 Chr.3: 49804291-49804291 on Build GRCh38
rs770649346 Chr.3: 49804216-49804216 on Build GRCh38
rs150931238 Chr.3: 49804209-49804209 on Build GRCh38
rs763314170 Chr.3: 49804257-49804257 on Build GRCh38
rs201114443 Chr.3: 49804297-49804297 on Build GRCh38
rs375329824 Chr.3: 49804168-49804168 on Build GRCh38
rs901071345 Chr.3: 49804330-49804330 on Build GRCh38
rs762419909 Chr.3: 49804240-49804240 on Build GRCh38
rs375293695 Chr.3: 49804252-49804252 on Build GRCh38
rs201597440 Chr.3: 49804227-49804227 on Build GRCh38
rs777691753 Chr.3: 49804196-49804196 on Build GRCh38
rs398091451 Chr.3: 49804136-49804136 on Build GRCh38
rs758683189 Chr.3: 49804156-49804156 on Build GRCh38
rs751805421 Chr.3: 49804279-49804279 on Build GRCh38
rs780945959 Chr.3: 49804174-49804174 on Build GRCh38
rs757149305 Chr.3: 49804322-49804322 on Build GRCh38
rs142871609 Chr.3: 49804170-49804170 on Build GRCh38
rs368340218 Chr.3: 49804298-49804298 on Build GRCh38
rs773681101 Chr.3: 49804255-49804255 on Build GRCh38
rs78612293 Chr.3: 49804135-49804135 on Build GRCh38
rs754633052 Chr.3: 49804110-49804110 on Build GRCh38

View Full Product Details