Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10065799-10066058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYS1
Entrez Gene ID
192668
Gene Name
cystin 1
Gene Aliases
-
UniGene
Hs.27092
Gene Chromosome Location
Chr.2: 10056799-10080411 on Build GRCh38
Transcripts
NM_001037160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960875 c.348C>A p.H116Q Chr.2: 10065927-10065927 on Build GRCh38
6639516 c.325G>A p.A109T Chr.2: 10065950-10065950 on Build GRCh38
3757539 c.357C>T p.S119S Chr.2: 10065918-10065918 on Build GRCh38
6266767 c.371+8C>A p.? Chr.2: 10065896-10065896 on Build GRCh38
6258810 c.326C>A p.A109E Chr.2: 10065949-10065949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558999780 Chr.2: 10066006-10066006 on Build GRCh38
rs768553125 Chr.2: 10065906-10065907 on Build GRCh38
rs775934549 Chr.2: 10065917-10065917 on Build GRCh38
rs142342337 Chr.2: 10065821-10065821 on Build GRCh38
rs763401516 Chr.2: 10065983-10065983 on Build GRCh38
rs778921847 Chr.2: 10065884-10065884 on Build GRCh38
rs936417353 Chr.2: 10065916-10065916 on Build GRCh38
rs750142139 Chr.2: 10065819-10065819 on Build GRCh38
rs917815876 Chr.2: 10065825-10065825 on Build GRCh38
rs570328700 Chr.2: 10065997-10065997 on Build GRCh38
rs182884229 Chr.2: 10065950-10065950 on Build GRCh38
rs776768606 Chr.2: 10065933-10065944 on Build GRCh38
rs755117187 Chr.2: 10065867-10065867 on Build GRCh38
rs375641931 Chr.2: 10065974-10065974 on Build GRCh38
rs548377123 Chr.2: 10065913-10065913 on Build GRCh38
rs373915269 Chr.2: 10065935-10065935 on Build GRCh38
rs374779271 Chr.2: 10065989-10065989 on Build GRCh38
rs377476624 Chr.2: 10065855-10065855 on Build GRCh38
rs539351229 Chr.2: 10066004-10066004 on Build GRCh38
rs145751380 Chr.2: 10065934-10065934 on Build GRCh38
rs745648295 Chr.2: 10065887-10065887 on Build GRCh38
rs768221819 Chr.2: 10065914-10065914 on Build GRCh38
rs750395519 Chr.2: 10066008-10066008 on Build GRCh38
rs777360684 Chr.2: 10065925-10065925 on Build GRCh38
rs767622899 Chr.2: 10066000-10066000 on Build GRCh38
rs200708612 Chr.2: 10065980-10065980 on Build GRCh38
rs969212936 Chr.2: 10065835-10065835 on Build GRCh38
rs1007507719 Chr.2: 10066038-10066038 on Build GRCh38
rs373526074 Chr.2: 10065866-10065866 on Build GRCh38
rs115484371 Chr.2: 10065924-10065924 on Build GRCh38
rs747634706 Chr.2: 10065968-10065968 on Build GRCh38
rs752822258 Chr.2: 10065948-10065948 on Build GRCh38
rs377581230 Chr.2: 10065918-10065918 on Build GRCh38
rs771904245 Chr.2: 10065903-10065903 on Build GRCh38
rs746663693 Chr.2: 10065951-10065951 on Build GRCh38
rs770927560 Chr.2: 10065880-10065880 on Build GRCh38
rs763271790 Chr.2: 10065928-10065928 on Build GRCh38
rs751830255 Chr.2: 10065933-10065933 on Build GRCh38
rs766613745 Chr.2: 10065929-10065929 on Build GRCh38
rs756790430 Chr.2: 10066039-10066039 on Build GRCh38
rs979639402 Chr.2: 10065921-10065921 on Build GRCh38
rs749264028 Chr.2: 10065877-10065877 on Build GRCh38
rs774513769 Chr.2: 10065991-10065991 on Build GRCh38
rs146537159 Chr.2: 10065960-10065960 on Build GRCh38
rs768187384 Chr.2: 10065966-10065966 on Build GRCh38
rs775234750 Chr.2: 10065909-10065909 on Build GRCh38
rs372362324 Chr.2: 10065963-10065963 on Build GRCh38

View Full Product Details