Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92837369-92837640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
-
UniGene
Hs.180946
Gene Chromosome Location
Chr.1: 92832729-92961522 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3 NR_000006.8

Gene Symbol
RPL5
Entrez Gene ID
6125
Gene Name
ribosomal protein L5
Gene Aliases
DBA6, L5, MSTP030, PPP1R135
UniGene
Hs.532359
Gene Chromosome Location
Chr.1: 92832037-92841924 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3 NR_000006.8

Gene Symbol
SNORD21
Entrez Gene ID
6083
Gene Name
small nucleolar RNA, C/D box 21
Gene Aliases
RNU21, U21
UniGene
Gene Chromosome Location
Chr.1: 92837289-92837383 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3 NR_000006.8

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
913349 c.666G>A p.Q222Q Chr.1: 92837594-92837594 on Build GRCh38
3736984 c.535C>T p.R179* Chr.1: 92837463-92837463 on Build GRCh38
913348 c.626G>A p.R209H Chr.1: 92837554-92837554 on Build GRCh38
4242895 c.541C>T p.P181S Chr.1: 92837469-92837469 on Build GRCh38
913347 c.614C>T p.A205V Chr.1: 92837542-92837542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750119521 Chr.1: 92837439-92837439 on Build GRCh38
rs966744388 Chr.1: 92837395-92837395 on Build GRCh38
rs371345041 Chr.1: 92837392-92837392 on Build GRCh38
rs369811492 Chr.1: 92837427-92837427 on Build GRCh38
rs767076160 Chr.1: 92837430-92837430 on Build GRCh38
rs985392468 Chr.1: 92837477-92837477 on Build GRCh38
rs965247191 Chr.1: 92837495-92837495 on Build GRCh38
rs779537310 Chr.1: 92837459-92837459 on Build GRCh38
rs749153960 Chr.1: 92837473-92837473 on Build GRCh38
rs754720452 Chr.1: 92837484-92837484 on Build GRCh38
rs587777118 Chr.1: 92837592-92837592 on Build GRCh38
rs778891633 Chr.1: 92837503-92837503 on Build GRCh38
rs764296257 Chr.1: 92837570-92837570 on Build GRCh38
rs762357870 Chr.1: 92837589-92837589 on Build GRCh38
rs749988750 Chr.1: 92837597-92837597 on Build GRCh38
rs760810833 Chr.1: 92837565-92837567 on Build GRCh38
rs931019369 Chr.1: 92837555-92837555 on Build GRCh38
rs546946459 Chr.1: 92837426-92837426 on Build GRCh38
rs776644546 Chr.1: 92837517-92837517 on Build GRCh38
rs11540832 Chr.1: 92837557-92837557 on Build GRCh38
rs755553750 Chr.1: 92837605-92837605 on Build GRCh38
rs375148060 Chr.1: 92837444-92837444 on Build GRCh38
rs763197787 Chr.1: 92837408-92837408 on Build GRCh38
rs753066236 Chr.1: 92837407-92837407 on Build GRCh38
rs756830726 Chr.1: 92837428-92837428 on Build GRCh38
rs767163981 Chr.1: 92837594-92837594 on Build GRCh38
rs770095159 Chr.1: 92837553-92837553 on Build GRCh38
rs764719541 Chr.1: 92837413-92837413 on Build GRCh38
rs773325598 Chr.1: 92837527-92837527 on Build GRCh38
rs775010708 Chr.1: 92837579-92837579 on Build GRCh38
rs771013653 Chr.1: 92837515-92837515 on Build GRCh38
rs914407234 Chr.1: 92837504-92837504 on Build GRCh38
rs752598659 Chr.1: 92837407-92837407 on Build GRCh38
rs746120360 Chr.1: 92837547-92837547 on Build GRCh38
rs528939210 Chr.1: 92837514-92837514 on Build GRCh38
rs775560628 Chr.1: 92837554-92837554 on Build GRCh38
rs765544108 Chr.1: 92837402-92837402 on Build GRCh38

View Full Product Details