Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43624713-43624938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COA1
Entrez Gene ID
55744
Gene Name
cytochrome c oxidase assembly factor 1 homolog
Gene Aliases
C7orf44, MITRAC15
UniGene
Hs.268887
Gene Chromosome Location
Chr.7: 43608452-43729541 on Build GRCh38
Transcripts
NR_135580.1 NR_135581.1 NR_135582.1 NR_135583.1 XM_005249807.4 XM_006715752.2 XM_017012414.1 XM_017012415.1 XM_017012416.1 XM_017012417.1 XM_017012418.1 XM_017012419.1 XM_017012421.1 XM_017012422.1 XM_017012424.1 XM_017012430.1 XM_017012431.1 XM_017012432.1 NM_004760.2 XM_017012792.1

Gene Symbol
STK17A
Entrez Gene ID
9263
Gene Name
serine/threonine kinase 17a
Gene Aliases
DRAK1
UniGene
Hs.709489
Gene Chromosome Location
Chr.7: 43583093-43627379 on Build GRCh38
Transcripts
NR_135580.1 NR_135581.1 NR_135582.1 NR_135583.1 XM_005249807.4 XM_006715752.2 XM_017012414.1 XM_017012415.1 XM_017012416.1 XM_017012417.1 XM_017012418.1 XM_017012419.1 XM_017012421.1 XM_017012422.1 XM_017012424.1 XM_017012430.1 XM_017012431.1 XM_017012432.1 NM_004760.2 XM_017012792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1089835 c.1235T>G p.F412C Chr.7: 43624832-43624832 on Build GRCh38
6286476 c.1238T>C p.I413T Chr.7: 43624835-43624835 on Build GRCh38
1089833 c.1135G>T p.G379* Chr.7: 43624732-43624732 on Build GRCh38
6471902 c.1239C>T p.I413I Chr.7: 43624836-43624836 on Build GRCh38
6791946 c.1223T>G p.I408S Chr.7: 43624820-43624820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019220298 Chr.7: 43624748-43624748 on Build GRCh38
rs375843224 Chr.7: 43624814-43624814 on Build GRCh38
rs144740791 Chr.7: 43624841-43624841 on Build GRCh38
rs753056472 Chr.7: 43624774-43624774 on Build GRCh38
rs1021128612 Chr.7: 43624833-43624833 on Build GRCh38
rs777009041 Chr.7: 43624764-43624764 on Build GRCh38
rs776523503 Chr.7: 43624860-43624860 on Build GRCh38
rs751758775 Chr.7: 43624782-43624782 on Build GRCh38
rs963663218 Chr.7: 43624799-43624799 on Build GRCh38
rs770257334 Chr.7: 43624866-43624866 on Build GRCh38
rs770814106 Chr.7: 43624831-43624833 on Build GRCh38
rs142571975 Chr.7: 43624838-43624838 on Build GRCh38
rs371595934 Chr.7: 43624768-43624768 on Build GRCh38
rs1006539588 Chr.7: 43624746-43624746 on Build GRCh38
rs781270039 Chr.7: 43624787-43624787 on Build GRCh38
rs781186005 Chr.7: 43624891-43624892 on Build GRCh38
rs775882759 Chr.7: 43624875-43624875 on Build GRCh38
rs969814970 Chr.7: 43624865-43624865 on Build GRCh38
rs766143001 Chr.7: 43624883-43624883 on Build GRCh38
rs747768998 Chr.7: 43624840-43624840 on Build GRCh38
rs780033239 Chr.7: 43624804-43624804 on Build GRCh38
rs756113243 Chr.7: 43624803-43624803 on Build GRCh38
rs142415211 Chr.7: 43624884-43624884 on Build GRCh38
rs150230872 Chr.7: 43624769-43624769 on Build GRCh38
rs138840285 Chr.7: 43624808-43624808 on Build GRCh38
rs762040230 Chr.7: 43624886-43624886 on Build GRCh38
rs773957244 Chr.7: 43624744-43624744 on Build GRCh38
rs749066768 Chr.7: 43624805-43624805 on Build GRCh38
rs193218605 Chr.7: 43624786-43624786 on Build GRCh38
rs764280193 Chr.7: 43624781-43624781 on Build GRCh38
rs183178602 Chr.7: 43624859-43624859 on Build GRCh38
rs761249710 Chr.7: 43624758-43624758 on Build GRCh38
rs201305620 Chr.7: 43624763-43624763 on Build GRCh38
rs15866 Chr.7: 43624878-43624878 on Build GRCh38
rs750480703 Chr.7: 43624893-43624893 on Build GRCh38
rs767557100 Chr.7: 43624890-43624890 on Build GRCh38
rs772766067 Chr.7: 43624850-43624850 on Build GRCh38
rs745847482 Chr.7: 43624797-43624797 on Build GRCh38

View Full Product Details