Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59938894-59939141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM217B
Entrez Gene ID
63939
Gene Name
family with sequence similarity 217 member B
Gene Aliases
C20orf177, dJ551D2.5
UniGene
Hs.54973
Gene Chromosome Location
Chr.20: 59933764-59948680 on Build GRCh38
Transcripts
NM_001190826.1 XM_011528986.2 NM_006242.3

Gene Symbol
PPP1R3D
Entrez Gene ID
5509
Gene Name
protein phosphatase 1 regulatory subunit 3D
Gene Aliases
PPP1R6
UniGene
Hs.42215
Gene Chromosome Location
Chr.20: 59936832-59940297 on Build GRCh38
Transcripts
NM_001190826.1 XM_011528986.2 NM_006242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4099900 c.792C>T p.A264A Chr.20: 59939140-59939140 on Build GRCh38
4099899 c.839G>A p.R280H Chr.20: 59939093-59939093 on Build GRCh38
2765464 c.797A>G p.Y266C Chr.20: 59939135-59939135 on Build GRCh38
6872294 c.795G>A p.E265E Chr.20: 59939137-59939137 on Build GRCh38
6224681 c.865G>A p.G289R Chr.20: 59939067-59939067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758454930 Chr.20: 59939010-59939010 on Build GRCh38
rs902008790 Chr.20: 59938970-59938970 on Build GRCh38
rs371051588 Chr.20: 59939121-59939121 on Build GRCh38
rs17854472 Chr.20: 59939103-59939103 on Build GRCh38
rs371149280 Chr.20: 59939015-59939015 on Build GRCh38
rs17854471 Chr.20: 59939115-59939115 on Build GRCh38
rs770503231 Chr.20: 59939025-59939025 on Build GRCh38
rs776027844 Chr.20: 59939033-59939033 on Build GRCh38
rs551576143 Chr.20: 59939053-59939053 on Build GRCh38
rs1042394154 Chr.20: 59939036-59939036 on Build GRCh38
rs750477897 Chr.20: 59939006-59939006 on Build GRCh38
rs777796623 Chr.20: 59939065-59939065 on Build GRCh38
rs745514874 Chr.20: 59939076-59939076 on Build GRCh38
rs768235278 Chr.20: 59939003-59939003 on Build GRCh38
rs774011048 Chr.20: 59939027-59939027 on Build GRCh38
rs774974410 Chr.20: 59939085-59939085 on Build GRCh38
rs761154540 Chr.20: 59939035-59939035 on Build GRCh38
rs780157665 Chr.20: 59939059-59939059 on Build GRCh38
rs750617695 Chr.20: 59939057-59939057 on Build GRCh38
rs756127577 Chr.20: 59939064-59939064 on Build GRCh38
rs778473475 Chr.20: 59939075-59939075 on Build GRCh38
rs769759075 Chr.20: 59939021-59939021 on Build GRCh38
rs757041020 Chr.20: 59939070-59939070 on Build GRCh38
rs139860582 Chr.20: 59939118-59939118 on Build GRCh38
rs997358557 Chr.20: 59939106-59939106 on Build GRCh38
rs771583006 Chr.20: 59939030-59939030 on Build GRCh38
rs769161377 Chr.20: 59939097-59939097 on Build GRCh38
rs779121626 Chr.20: 59939005-59939005 on Build GRCh38
rs746447259 Chr.20: 59939096-59939096 on Build GRCh38
rs749025787 Chr.20: 59939024-59939024 on Build GRCh38
rs771543397 Chr.20: 59939077-59939077 on Build GRCh38
rs371403831 Chr.20: 59939042-59939042 on Build GRCh38
rs556641157 Chr.20: 59938960-59938960 on Build GRCh38
rs748065432 Chr.20: 59939017-59939017 on Build GRCh38
rs1040533448 Chr.20: 59938950-59938950 on Build GRCh38
rs376738312 Chr.20: 59939089-59939089 on Build GRCh38
rs757463070 Chr.20: 59938999-59938999 on Build GRCh38
rs200816379 Chr.20: 59938987-59938987 on Build GRCh38
rs567015173 Chr.20: 59939067-59939067 on Build GRCh38
rs777707019 Chr.20: 59939022-59939022 on Build GRCh38
rs758446264 Chr.20: 59939058-59939058 on Build GRCh38
rs759003138 Chr.20: 59939029-59939029 on Build GRCh38
rs762116063 Chr.20: 59939112-59939112 on Build GRCh38
rs773353751 Chr.20: 59939119-59939119 on Build GRCh38
rs754202362 Chr.20: 59939050-59939050 on Build GRCh38
rs765255477 Chr.20: 59938982-59938982 on Build GRCh38

View Full Product Details