Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27099472-27099697 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGREF1
Entrez Gene ID
10669
Gene Name
cell growth regulator with EF-hand domain 1
Gene Aliases
CGR11
UniGene
Hs.159525
Gene Chromosome Location
Chr.2: 27099353-27119127 on Build GRCh38
Transcripts
NM_001166240.1 NM_001301324.1 NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_017004060.1 XM_017004061.1

Gene Symbol
KHK
Entrez Gene ID
3795
Gene Name
ketohexokinase
Gene Aliases
-
UniGene
Hs.567297
Gene Chromosome Location
Chr.2: 27086738-27100751 on Build GRCh38
Transcripts
NM_001166240.1 NM_001301324.1 NM_000221.2 NM_006488.2 XM_005264294.3 XM_005264296.3 XM_005264298.3 XM_017004060.1 XM_017004061.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3991239 c.733T>C p.F245L Chr.2: 27099499-27099499 on Build GRCh38
1165924 c.815G>A p.R272K Chr.2: 27099668-27099668 on Build GRCh38
1019515 c.819C>T p.S273S Chr.2: 27099672-27099672 on Build GRCh38
6835519 c.723C>G p.H241Q Chr.2: 27099489-27099489 on Build GRCh38
3839250 c.745C>T p.R249C Chr.2: 27099511-27099511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760617762 Chr.2: 27099590-27099590 on Build GRCh38
rs770569236 Chr.2: 27099653-27099653 on Build GRCh38
rs780112409 Chr.2: 27099536-27099536 on Build GRCh38
rs1033932441 Chr.2: 27099599-27099599 on Build GRCh38
rs61735560 Chr.2: 27099670-27099670 on Build GRCh38
rs747498412 Chr.2: 27099540-27099540 on Build GRCh38
rs748640362 Chr.2: 27099624-27099624 on Build GRCh38
rs770353880 Chr.2: 27099558-27099558 on Build GRCh38
rs959830044 Chr.2: 27099611-27099611 on Build GRCh38
rs918025823 Chr.2: 27099626-27099626 on Build GRCh38
rs776433978 Chr.2: 27099677-27099677 on Build GRCh38
rs549772974 Chr.2: 27099512-27099512 on Build GRCh38
rs1052033257 Chr.2: 27099569-27099569 on Build GRCh38
rs372381680 Chr.2: 27099605-27099605 on Build GRCh38
rs767615753 Chr.2: 27099580-27099580 on Build GRCh38
rs199989985 Chr.2: 27099511-27099511 on Build GRCh38
rs781648272 Chr.2: 27099548-27099548 on Build GRCh38
rs773873126 Chr.2: 27099559-27099559 on Build GRCh38
rs114353144 Chr.2: 27099556-27099556 on Build GRCh38
rs138164728 Chr.2: 27099673-27099673 on Build GRCh38
rs770138704 Chr.2: 27099553-27099553 on Build GRCh38
rs770159249 Chr.2: 27099642-27099642 on Build GRCh38
rs7578864 Chr.2: 27099582-27099582 on Build GRCh38
rs776125990 Chr.2: 27099492-27099492 on Build GRCh38
rs144520325 Chr.2: 27099492-27099492 on Build GRCh38
rs780015909 Chr.2: 27099607-27099607 on Build GRCh38
rs751705633 Chr.2: 27099612-27099612 on Build GRCh38
rs752495277 Chr.2: 27099513-27099513 on Build GRCh38
rs371516742 Chr.2: 27099520-27099520 on Build GRCh38
rs748925154 Chr.2: 27099650-27099650 on Build GRCh38
rs940048853 Chr.2: 27099630-27099630 on Build GRCh38
rs202164115 Chr.2: 27099577-27099577 on Build GRCh38
rs781712588 Chr.2: 27099623-27099623 on Build GRCh38
rs141220653 Chr.2: 27099535-27099535 on Build GRCh38
rs751466422 Chr.2: 27099504-27099504 on Build GRCh38
rs771991153 Chr.2: 27099555-27099555 on Build GRCh38
rs773856508 Chr.2: 27099656-27099656 on Build GRCh38
rs774520724 Chr.2: 27099538-27099538 on Build GRCh38
rs750612355 Chr.2: 27099604-27099604 on Build GRCh38
rs778134637 Chr.2: 27099649-27099649 on Build GRCh38
rs144943949 Chr.2: 27099552-27099552 on Build GRCh38
rs368056714 Chr.2: 27099503-27099503 on Build GRCh38
rs769098063 Chr.2: 27099543-27099543 on Build GRCh38
rs1005052731 Chr.2: 27099636-27099636 on Build GRCh38
rs755019513 Chr.2: 27099613-27099613 on Build GRCh38
rs755955022 Chr.2: 27099514-27099514 on Build GRCh38
rs201072096 Chr.2: 27099594-27099594 on Build GRCh38
rs758419401 Chr.2: 27099525-27099525 on Build GRCh38
rs575951862 Chr.2: 27099671-27099671 on Build GRCh38
rs543317237 Chr.2: 27099672-27099672 on Build GRCh38

View Full Product Details