Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59939300-59939533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM217B
Entrez Gene ID
63939
Gene Name
family with sequence similarity 217 member B
Gene Aliases
C20orf177, dJ551D2.5
UniGene
Hs.54973
Gene Chromosome Location
Chr.20: 59933764-59948680 on Build GRCh38
Transcripts
NM_001190826.1 XM_011528986.2 NM_006242.3

Gene Symbol
PPP1R3D
Entrez Gene ID
5509
Gene Name
protein phosphatase 1 regulatory subunit 3D
Gene Aliases
PPP1R6
UniGene
Hs.42215
Gene Chromosome Location
Chr.20: 59936832-59940297 on Build GRCh38
Transcripts
NM_001190826.1 XM_011528986.2 NM_006242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5366014 c.477C>T p.F159F Chr.20: 59939455-59939455 on Build GRCh38
5462981 c.483G>A p.P161P Chr.20: 59939449-59939449 on Build GRCh38
5474294 c.619C>T p.R207C Chr.20: 59939313-59939313 on Build GRCh38
2765478 c.580C>T p.R194C Chr.20: 59939352-59939352 on Build GRCh38
2765476 c.609G>A p.Q203Q Chr.20: 59939323-59939323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376633935 Chr.20: 59939477-59939477 on Build GRCh38
rs372312575 Chr.20: 59939327-59939327 on Build GRCh38
rs776378402 Chr.20: 59939509-59939509 on Build GRCh38
rs774322736 Chr.20: 59939481-59939481 on Build GRCh38
rs750924836 Chr.20: 59939452-59939452 on Build GRCh38
rs755702252 Chr.20: 59939347-59939347 on Build GRCh38
rs957491895 Chr.20: 59939428-59939428 on Build GRCh38
rs761446726 Chr.20: 59939514-59939514 on Build GRCh38
rs760464270 Chr.20: 59939504-59939504 on Build GRCh38
rs539093057 Chr.20: 59939376-59939376 on Build GRCh38
rs147721457 Chr.20: 59939390-59939390 on Build GRCh38
rs745793631 Chr.20: 59939486-59939486 on Build GRCh38
rs770024453 Chr.20: 59939462-59939468 on Build GRCh38
rs754465409 Chr.20: 59939362-59939362 on Build GRCh38
rs555670934 Chr.20: 59939490-59939490 on Build GRCh38
rs780359967 Chr.20: 59939389-59939389 on Build GRCh38
rs780267621 Chr.20: 59939455-59939455 on Build GRCh38
rs888453640 Chr.20: 59939488-59939488 on Build GRCh38
rs758843522 Chr.20: 59939453-59939453 on Build GRCh38
rs1050573083 Chr.20: 59939482-59939482 on Build GRCh38
rs767195432 Chr.20: 59939430-59939430 on Build GRCh38
rs763673526 Chr.20: 59939506-59939506 on Build GRCh38
rs763737797 Chr.20: 59939437-59939437 on Build GRCh38
rs767231974 Chr.20: 59939336-59939336 on Build GRCh38
rs980601297 Chr.20: 59939398-59939398 on Build GRCh38
rs766226789 Chr.20: 59939331-59939331 on Build GRCh38
rs752260687 Chr.20: 59939346-59939346 on Build GRCh38
rs557669925 Chr.20: 59939433-59939433 on Build GRCh38
rs746281614 Chr.20: 59939399-59939426 on Build GRCh38
rs768838779 Chr.20: 59939403-59939403 on Build GRCh38
rs747322374 Chr.20: 59939458-59939458 on Build GRCh38
rs946038649 Chr.20: 59939447-59939447 on Build GRCh38
rs751359121 Chr.20: 59939332-59939332 on Build GRCh38
rs756775722 Chr.20: 59939443-59939443 on Build GRCh38
rs923532419 Chr.20: 59939462-59939462 on Build GRCh38
rs775157317 Chr.20: 59939494-59939494 on Build GRCh38
rs149736385 Chr.20: 59939439-59939439 on Build GRCh38
rs773249476 Chr.20: 59939320-59939320 on Build GRCh38
rs781332810 Chr.20: 59939466-59939466 on Build GRCh38
rs142381631 Chr.20: 59939411-59939411 on Build GRCh38
rs769836216 Chr.20: 59939471-59939471 on Build GRCh38
rs774234822 Chr.20: 59939421-59939421 on Build GRCh38
rs758645982 Chr.20: 59939387-59939387 on Build GRCh38
rs540042164 Chr.20: 59939446-59939446 on Build GRCh38
rs571783929 Chr.20: 59939368-59939368 on Build GRCh38
rs570881754 Chr.20: 59939378-59939378 on Build GRCh38
rs755158732 Chr.20: 59939461-59939461 on Build GRCh38
rs748260351 Chr.20: 59939467-59939467 on Build GRCh38
rs763503005 Chr.20: 59939356-59939356 on Build GRCh38
rs776567247 Chr.20: 59939406-59939406 on Build GRCh38
rs759366185 Chr.20: 59939424-59939424 on Build GRCh38

View Full Product Details