Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116760768-116761025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUD13
Entrez Gene ID
84811
Gene Name
BUD13 homolog
Gene Aliases
Cwc26, fSAP71
UniGene
Hs.437341
Gene Chromosome Location
Chr.11: 116748170-116772998 on Build GRCh38
Transcripts
NM_001159736.1 NM_032725.3 XM_011543035.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357880 c.1163C>G p.S388C Chr.11: 116760826-116760826 on Build GRCh38
4018057 c.1079G>A p.R360Q Chr.11: 116760910-116760910 on Build GRCh38
4018056 c.1150C>T p.R384W Chr.11: 116760839-116760839 on Build GRCh38
3675985 c.1218G>C p.L406L Chr.11: 116760771-116760771 on Build GRCh38
4458148 c.1073C>T p.S358F Chr.11: 116760916-116760916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891862436 Chr.11: 116760895-116760895 on Build GRCh38
rs769257470 Chr.11: 116760959-116760959 on Build GRCh38
rs866623609 Chr.11: 116760812-116760812 on Build GRCh38
rs748247607 Chr.11: 116760805-116760805 on Build GRCh38
rs764209410 Chr.11: 116760995-116760995 on Build GRCh38
rs749542207 Chr.11: 116760820-116760820 on Build GRCh38
rs768156860 Chr.11: 116760812-116760818 on Build GRCh38
rs149005278 Chr.11: 116760839-116760839 on Build GRCh38
rs779061716 Chr.11: 116760930-116760930 on Build GRCh38
rs755516486 Chr.11: 116760793-116760793 on Build GRCh38
rs371950031 Chr.11: 116760881-116760881 on Build GRCh38
rs759916257 Chr.11: 116760968-116760968 on Build GRCh38
rs142510693 Chr.11: 116760902-116760902 on Build GRCh38
rs35004487 Chr.11: 116760826-116760826 on Build GRCh38
rs768373463 Chr.11: 116760946-116760946 on Build GRCh38
rs113542136 Chr.11: 116760838-116760838 on Build GRCh38
rs750032557 Chr.11: 116760857-116760857 on Build GRCh38
rs3825041 Chr.11: 116760991-116760991 on Build GRCh38
rs760471044 Chr.11: 116760828-116760828 on Build GRCh38
rs983232059 Chr.11: 116760836-116760836 on Build GRCh38
rs773944121 Chr.11: 116760966-116760966 on Build GRCh38
rs1054580991 Chr.11: 116760889-116760889 on Build GRCh38
rs781078672 Chr.11: 116760794-116760794 on Build GRCh38
rs778028939 Chr.11: 116760907-116760907 on Build GRCh38
rs372663182 Chr.11: 116760960-116760960 on Build GRCh38
rs374131301 Chr.11: 116760813-116760813 on Build GRCh38
rs946254909 Chr.11: 116760909-116760909 on Build GRCh38
rs757564436 Chr.11: 116760914-116760914 on Build GRCh38
rs1045904083 Chr.11: 116760922-116760922 on Build GRCh38
rs781721564 Chr.11: 116760894-116760894 on Build GRCh38
rs778110700 Chr.11: 116760819-116760819 on Build GRCh38
rs1017684654 Chr.11: 116760910-116760910 on Build GRCh38
rs368705815 Chr.11: 116760958-116760958 on Build GRCh38
rs189942409 Chr.11: 116760911-116760911 on Build GRCh38
rs746812727 Chr.11: 116760931-116760931 on Build GRCh38
rs761575761 Chr.11: 116760973-116760974 on Build GRCh38
rs3741300 Chr.11: 116760974-116760974 on Build GRCh38
rs151084467 Chr.11: 116760884-116760884 on Build GRCh38
rs766043963 Chr.11: 116760963-116760963 on Build GRCh38
rs771953293 Chr.11: 116760821-116760821 on Build GRCh38
rs761277212 Chr.11: 116760842-116760842 on Build GRCh38
rs776372936 Chr.11: 116760898-116760899 on Build GRCh38
rs753214569 Chr.11: 116760989-116760989 on Build GRCh38
rs184331046 Chr.11: 116760883-116760883 on Build GRCh38
rs762536948 Chr.11: 116760961-116760961 on Build GRCh38
rs148467711 Chr.11: 116760845-116760845 on Build GRCh38
rs753184596 Chr.11: 116760900-116760900 on Build GRCh38

View Full Product Details