Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27576945-27577216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf16
Entrez Gene ID
84226
Gene Name
chromosome 2 open reading frame 16
Gene Aliases
-
UniGene
Hs.131021
Gene Chromosome Location
Chr.2: 27576522-27582722 on Build GRCh38
Transcripts
NM_032266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019741 c.420G>T p.K140N Chr.2: 27576992-27576992 on Build GRCh38
1019745 c.533C>A p.S178Y Chr.2: 27577105-27577105 on Build GRCh38
7352703 c.497G>T p.G166V Chr.2: 27577069-27577069 on Build GRCh38
5127795 c.404_407delTGAT p.M135fs*14 Chr.2: 27576976-27576979 on Build GRCh38
1019743 c.426A>C p.Q142H Chr.2: 27576998-27576998 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903861843 Chr.2: 27576989-27576989 on Build GRCh38
rs866490864 Chr.2: 27577077-27577077 on Build GRCh38
rs770841593 Chr.2: 27576979-27576979 on Build GRCh38
rs888152474 Chr.2: 27577091-27577091 on Build GRCh38
rs749558119 Chr.2: 27576978-27576978 on Build GRCh38
rs865983082 Chr.2: 27577153-27577153 on Build GRCh38
rs774821958 Chr.2: 27576996-27576996 on Build GRCh38
rs769494851 Chr.2: 27577179-27577179 on Build GRCh38
rs780019428 Chr.2: 27577133-27577133 on Build GRCh38
rs745732845 Chr.2: 27576983-27576983 on Build GRCh38
rs761522359 Chr.2: 27577024-27577024 on Build GRCh38
rs768151063 Chr.2: 27577147-27577147 on Build GRCh38
rs79989314 Chr.2: 27577040-27577040 on Build GRCh38
rs752779376 Chr.2: 27577063-27577063 on Build GRCh38
rs180785122 Chr.2: 27577069-27577069 on Build GRCh38
rs747742925 Chr.2: 27577170-27577170 on Build GRCh38
rs778167714 Chr.2: 27576968-27576968 on Build GRCh38
rs776235165 Chr.2: 27577019-27577019 on Build GRCh38
rs778503939 Chr.2: 27577108-27577108 on Build GRCh38
rs536164421 Chr.2: 27577017-27577017 on Build GRCh38
rs571976158 Chr.2: 27576974-27576974 on Build GRCh38
rs760199824 Chr.2: 27577002-27577002 on Build GRCh38
rs760911943 Chr.2: 27577087-27577087 on Build GRCh38
rs1025599718 Chr.2: 27577096-27577096 on Build GRCh38
rs766642400 Chr.2: 27577028-27577028 on Build GRCh38
rs757511771 Chr.2: 27577107-27577107 on Build GRCh38
rs778823292 Chr.2: 27576981-27576981 on Build GRCh38
rs772049121 Chr.2: 27576992-27576992 on Build GRCh38
rs759896889 Chr.2: 27577041-27577041 on Build GRCh38
rs746988640 Chr.2: 27577135-27577135 on Build GRCh38
rs776179241 Chr.2: 27577150-27577150 on Build GRCh38
rs745705052 Chr.2: 27577111-27577111 on Build GRCh38
rs764590022 Chr.2: 27577137-27577137 on Build GRCh38
rs768104906 Chr.2: 27577061-27577061 on Build GRCh38
rs759971498 Chr.2: 27577186-27577186 on Build GRCh38
rs778061664 Chr.2: 27577075-27577075 on Build GRCh38
rs544686945 Chr.2: 27577129-27577129 on Build GRCh38
rs754097355 Chr.2: 27577080-27577080 on Build GRCh38
rs774542360 Chr.2: 27577182-27577182 on Build GRCh38
rs777465948 Chr.2: 27577164-27577165 on Build GRCh38

View Full Product Details