Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93562238-93562465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAR3
Entrez Gene ID
8412
Gene Name
breast cancer anti-estrogen resistance 3
Gene Aliases
NSP2, SH2D3B
UniGene
Hs.36958
Gene Chromosome Location
Chr.1: 93561741-93847150 on Build GRCh38
Transcripts
NM_001261408.1 NM_001261409.1 NM_001261410.1 NM_001308251.1 NM_003567.3 XM_011542251.2 XM_017002480.1 XM_011541625.2

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
UniGene
Hs.134060
Gene Chromosome Location
Chr.1: 93448067-93569671 on Build GRCh38
Transcripts
NM_001261408.1 NM_001261409.1 NM_001261410.1 NM_001308251.1 NM_003567.3 XM_011542251.2 XM_017002480.1 XM_011541625.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3790352 c.2085G>C p.L695F Chr.1: 93562361-93562361 on Build GRCh38
374928 c.2360T>C p.L787P Chr.1: 93562359-93562359 on Build GRCh38
3790351 c.2358G>C p.L786F Chr.1: 93562361-93562361 on Build GRCh38
913405 c.2438G>A p.R813H Chr.1: 93562281-93562281 on Build GRCh38
913406 c.2311G>A p.D771N Chr.1: 93562408-93562408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775633367 Chr.1: 93562425-93562425 on Build GRCh38
rs547841302 Chr.1: 93562270-93562270 on Build GRCh38
rs780958961 Chr.1: 93562264-93562264 on Build GRCh38
rs756435162 Chr.1: 93562435-93562438 on Build GRCh38
rs768510746 Chr.1: 93562383-93562383 on Build GRCh38
rs1019912706 Chr.1: 93562376-93562376 on Build GRCh38
rs965616097 Chr.1: 93562397-93562397 on Build GRCh38
rs150697615 Chr.1: 93562396-93562396 on Build GRCh38
rs759562164 Chr.1: 93562405-93562405 on Build GRCh38
rs755856869 Chr.1: 93562363-93562363 on Build GRCh38
rs748027776 Chr.1: 93562388-93562388 on Build GRCh38
rs144562250 Chr.1: 93562315-93562315 on Build GRCh38
rs769329876 Chr.1: 93562301-93562301 on Build GRCh38
rs750142598 Chr.1: 93562360-93562360 on Build GRCh38
rs764599323 Chr.1: 93562350-93562350 on Build GRCh38
rs763367246 Chr.1: 93562434-93562434 on Build GRCh38
rs866202593 Chr.1: 93562308-93562308 on Build GRCh38
rs373868471 Chr.1: 93562281-93562281 on Build GRCh38
rs149846383 Chr.1: 93562410-93562410 on Build GRCh38
rs779816516 Chr.1: 93562365-93562365 on Build GRCh38
rs776977845 Chr.1: 93562305-93562305 on Build GRCh38
rs778964260 Chr.1: 93562387-93562387 on Build GRCh38
rs769751257 Chr.1: 93562271-93562271 on Build GRCh38
rs147155903 Chr.1: 93562391-93562391 on Build GRCh38
rs770901795 Chr.1: 93562288-93562288 on Build GRCh38
rs751998658 Chr.1: 93562352-93562352 on Build GRCh38
rs749096646 Chr.1: 93562366-93562366 on Build GRCh38
rs755217618 Chr.1: 93562262-93562264 on Build GRCh38
rs767707091 Chr.1: 93562346-93562346 on Build GRCh38
rs763261459 Chr.1: 93562332-93562332 on Build GRCh38
rs367823753 Chr.1: 93562282-93562282 on Build GRCh38
rs369490013 Chr.1: 93562284-93562284 on Build GRCh38
rs753148375 Chr.1: 93562378-93562379 on Build GRCh38
rs143568257 Chr.1: 93562353-93562353 on Build GRCh38
rs150172728 Chr.1: 93562283-93562283 on Build GRCh38
rs759641595 Chr.1: 93562308-93562308 on Build GRCh38
rs753081234 Chr.1: 93562318-93562318 on Build GRCh38
rs371840059 Chr.1: 93562358-93562358 on Build GRCh38
rs997152293 Chr.1: 93562276-93562276 on Build GRCh38

View Full Product Details