Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 48693589-48693820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNT1
Entrez Gene ID
904
Gene Name
cyclin T1
Gene Aliases
CCNT, CYCT1, HIVE1
UniGene
Hs.92308
Gene Chromosome Location
Chr.12: 48688458-48716998 on Build GRCh38
Transcripts
NM_001240.3 NM_001277842.1 XM_017020197.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623101 c.1610C>T p.T537I Chr.12: 48693604-48693604 on Build GRCh38
1730402 c.1548T>G p.N516K Chr.12: 48693666-48693666 on Build GRCh38
5632686 c.1490A>G p.E497G Chr.12: 48693724-48693724 on Build GRCh38
6136875 c.1406G>A p.G469E Chr.12: 48693808-48693808 on Build GRCh38
4405963 c.1512A>G p.R504R Chr.12: 48693702-48693702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780374398 Chr.12: 48693792-48693792 on Build GRCh38
rs918870860 Chr.12: 48693738-48693738 on Build GRCh38
rs747030494 Chr.12: 48693677-48693677 on Build GRCh38
rs558225201 Chr.12: 48693745-48693745 on Build GRCh38
rs766286862 Chr.12: 48693669-48693669 on Build GRCh38
rs775752617 Chr.12: 48693698-48693698 on Build GRCh38
rs949231264 Chr.12: 48693627-48693627 on Build GRCh38
rs776007780 Chr.12: 48693612-48693612 on Build GRCh38
rs370391951 Chr.12: 48693675-48693675 on Build GRCh38
rs142303037 Chr.12: 48693658-48693658 on Build GRCh38
rs61755289 Chr.12: 48693795-48693795 on Build GRCh38
rs889822518 Chr.12: 48693680-48693680 on Build GRCh38
rs373807143 Chr.12: 48693685-48693685 on Build GRCh38
rs777859495 Chr.12: 48693676-48693676 on Build GRCh38
rs767245574 Chr.12: 48693653-48693653 on Build GRCh38
rs760407470 Chr.12: 48693728-48693728 on Build GRCh38
rs757269550 Chr.12: 48693766-48693766 on Build GRCh38
rs371197465 Chr.12: 48693651-48693653 on Build GRCh38
rs750164387 Chr.12: 48693656-48693656 on Build GRCh38
rs773758545 Chr.12: 48693630-48693630 on Build GRCh38
rs772937692 Chr.12: 48693719-48693719 on Build GRCh38
rs151213150 Chr.12: 48693748-48693748 on Build GRCh38
rs763623277 Chr.12: 48693621-48693621 on Build GRCh38
rs746054253 Chr.12: 48693777-48693777 on Build GRCh38
rs202226049 Chr.12: 48693769-48693769 on Build GRCh38
rs770202413 Chr.12: 48693611-48693611 on Build GRCh38
rs774960495 Chr.12: 48693712-48693712 on Build GRCh38
rs761399979 Chr.12: 48693716-48693716 on Build GRCh38
rs756045525 Chr.12: 48693781-48693781 on Build GRCh38
rs761461051 Chr.12: 48693639-48693639 on Build GRCh38
rs753595809 Chr.12: 48693740-48693740 on Build GRCh38
rs759891367 Chr.12: 48693610-48693610 on Build GRCh38
rs201423333 Chr.12: 48693796-48693796 on Build GRCh38
rs368324686 Chr.12: 48693690-48693690 on Build GRCh38
rs763378412 Chr.12: 48693613-48693613 on Build GRCh38
rs769023522 Chr.12: 48693704-48693704 on Build GRCh38
rs771638484 Chr.12: 48693799-48693799 on Build GRCh38
rs766158043 Chr.12: 48693734-48693734 on Build GRCh38
rs777826980 Chr.12: 48693783-48693810 on Build GRCh38
rs754850196 Chr.12: 48693744-48693744 on Build GRCh38
rs752739512 Chr.12: 48693756-48693756 on Build GRCh38
rs756355960 Chr.12: 48693782-48693782 on Build GRCh38
rs749756750 Chr.12: 48693703-48693703 on Build GRCh38
rs767190434 Chr.12: 48693718-48693718 on Build GRCh38
rs753742785 Chr.12: 48693670-48693670 on Build GRCh38
rs757395667 Chr.12: 48693682-48693682 on Build GRCh38
rs781254287 Chr.12: 48693767-48693767 on Build GRCh38
rs746103551 Chr.12: 48693688-48693688 on Build GRCh38

View Full Product Details