Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67162808-67163049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL8
Entrez Gene ID
55336
Gene Name
F-box and leucine rich repeat protein 8
Gene Aliases
FBL8
UniGene
Hs.710714
Gene Chromosome Location
Chr.16: 67159988-67164174 on Build GRCh38
Transcripts
NM_018378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4591721 c.193T>C p.S65P Chr.16: 67162888-67162888 on Build GRCh38
972297 c.319C>T p.R107C Chr.16: 67163014-67163014 on Build GRCh38
4591225 c.196G>C p.A66P Chr.16: 67162891-67162891 on Build GRCh38
3387532 c.260G>A p.R87Q Chr.16: 67162955-67162955 on Build GRCh38
4995674 c.220C>G p.L74V Chr.16: 67162915-67162915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754676264 Chr.16: 67162838-67162838 on Build GRCh38
rs945020865 Chr.16: 67162999-67162999 on Build GRCh38
rs972835072 Chr.16: 67162861-67162861 on Build GRCh38
rs375830131 Chr.16: 67163002-67163002 on Build GRCh38
rs774177986 Chr.16: 67162976-67162976 on Build GRCh38
rs747117718 Chr.16: 67162914-67162914 on Build GRCh38
rs758997725 Chr.16: 67162864-67162864 on Build GRCh38
rs371579014 Chr.16: 67162841-67162842 on Build GRCh38
rs866550676 Chr.16: 67162945-67162945 on Build GRCh38
rs772025378 Chr.16: 67162985-67162985 on Build GRCh38
rs765875636 Chr.16: 67162995-67162995 on Build GRCh38
rs748504461 Chr.16: 67162862-67162862 on Build GRCh38
rs764936780 Chr.16: 67163007-67163007 on Build GRCh38
rs551094669 Chr.16: 67162984-67162984 on Build GRCh38
rs903569211 Chr.16: 67163029-67163029 on Build GRCh38
rs865967857 Chr.16: 67162977-67162977 on Build GRCh38
rs753145061 Chr.16: 67163008-67163008 on Build GRCh38
rs1037130802 Chr.16: 67162886-67162886 on Build GRCh38
rs889942286 Chr.16: 67162968-67162968 on Build GRCh38
rs531379721 Chr.16: 67162975-67162975 on Build GRCh38
rs746141895 Chr.16: 67162955-67162955 on Build GRCh38
rs760652240 Chr.16: 67162991-67162991 on Build GRCh38
rs778103337 Chr.16: 67162891-67162891 on Build GRCh38
rs901231990 Chr.16: 67162895-67162895 on Build GRCh38
rs753360725 Chr.16: 67162998-67162998 on Build GRCh38
rs779380843 Chr.16: 67162952-67162952 on Build GRCh38
rs980259485 Chr.16: 67162913-67162913 on Build GRCh38
rs938975182 Chr.16: 67162954-67162954 on Build GRCh38
rs770858768 Chr.16: 67162947-67162947 on Build GRCh38
rs145203594 Chr.16: 67162965-67162965 on Build GRCh38
rs909907509 Chr.16: 67162833-67162833 on Build GRCh38
rs772952446 Chr.16: 67162987-67162987 on Build GRCh38
rs779462798 Chr.16: 67162847-67162847 on Build GRCh38
rs748115707 Chr.16: 67162903-67162904 on Build GRCh38
rs200909842 Chr.16: 67163019-67163019 on Build GRCh38

View Full Product Details