Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48022123-48022372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELSPBP1
Entrez Gene ID
64100
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12, EDDM12, EL149, HE12
UniGene
Hs.104894
Gene Chromosome Location
Chr.19: 47994651-48025153 on Build GRCh38
Transcripts
NM_022142.4 XM_017027130.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6558068 c.515G>T p.R172I Chr.19: 48022170-48022170 on Build GRCh38
998835 c.537C>A p.G179G Chr.19: 48022192-48022192 on Build GRCh38
5916716 c.515G>A p.R172K Chr.19: 48022170-48022170 on Build GRCh38
5894016 c.541C>T p.P181S Chr.19: 48022196-48022196 on Build GRCh38
7097076 c.645C>T p.D215D Chr.19: 48022300-48022300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757381004 Chr.19: 48022172-48022172 on Build GRCh38
rs6509358 Chr.19: 48022298-48022298 on Build GRCh38
rs201457821 Chr.19: 48022178-48022178 on Build GRCh38
rs767512692 Chr.19: 48022146-48022146 on Build GRCh38
rs769136306 Chr.19: 48022183-48022183 on Build GRCh38
rs112458959 Chr.19: 48022201-48022201 on Build GRCh38
rs759153571 Chr.19: 48022312-48022312 on Build GRCh38
rs779511106 Chr.19: 48022249-48022249 on Build GRCh38
rs754644214 Chr.19: 48022334-48022334 on Build GRCh38
rs532447692 Chr.19: 48022321-48022321 on Build GRCh38
rs780388434 Chr.19: 48022257-48022257 on Build GRCh38
rs773990420 Chr.19: 48022210-48022210 on Build GRCh38
rs778863071 Chr.19: 48022267-48022267 on Build GRCh38
rs748686790 Chr.19: 48022272-48022272 on Build GRCh38
rs780292149 Chr.19: 48022180-48022180 on Build GRCh38
rs770278847 Chr.19: 48022273-48022273 on Build GRCh38
rs777154310 Chr.19: 48022282-48022282 on Build GRCh38
rs953765953 Chr.19: 48022339-48022339 on Build GRCh38
rs373312661 Chr.19: 48022267-48022267 on Build GRCh38
rs113159670 Chr.19: 48022208-48022208 on Build GRCh38
rs769227693 Chr.19: 48022204-48022204 on Build GRCh38
rs769460296 Chr.19: 48022301-48022301 on Build GRCh38
rs764028805 Chr.19: 48022329-48022329 on Build GRCh38
rs752674666 Chr.19: 48022226-48022226 on Build GRCh38
rs2303690 Chr.19: 48022250-48022250 on Build GRCh38
rs75838973 Chr.19: 48022212-48022212 on Build GRCh38
rs138605860 Chr.19: 48022269-48022269 on Build GRCh38
rs149319517 Chr.19: 48022297-48022297 on Build GRCh38
rs774037686 Chr.19: 48022304-48022304 on Build GRCh38
rs751816896 Chr.19: 48022259-48022259 on Build GRCh38
rs767281706 Chr.19: 48022225-48022225 on Build GRCh38
rs1029204716 Chr.19: 48022317-48022317 on Build GRCh38
rs779023687 Chr.19: 48022177-48022177 on Build GRCh38
rs754939708 Chr.19: 48022193-48022193 on Build GRCh38
rs777334602 Chr.19: 48022189-48022189 on Build GRCh38
rs758594292 Chr.19: 48022179-48022179 on Build GRCh38
rs775221571 Chr.19: 48022316-48022316 on Build GRCh38
rs760569996 Chr.19: 48022244-48022244 on Build GRCh38
rs188877655 Chr.19: 48022154-48022154 on Build GRCh38
rs141592425 Chr.19: 48022206-48022206 on Build GRCh38
rs758550566 Chr.19: 48022254-48022254 on Build GRCh38
rs35700000 Chr.19: 48022209-48022209 on Build GRCh38

View Full Product Details