Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 72098964-72099231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX38
Entrez Gene ID
9785
Gene Name
DEAH-box helicase 38
Gene Aliases
DDX38, PRP16, PRPF16
UniGene
Hs.151218
Gene Chromosome Location
Chr.16: 72093665-72112912 on Build GRCh38
Transcripts
NM_014003.3 XM_005256269.1 XM_011523484.1 XM_011523485.1 XM_017023913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392223 c.831G>T p.W277C Chr.16: 72098993-72098993 on Build GRCh38
4509668 c.813C>T p.S271S Chr.16: 72098975-72098975 on Build GRCh38
6643659 c.826G>A p.E276K Chr.16: 72098988-72098988 on Build GRCh38
3969685 c.863C>T p.P288L Chr.16: 72099025-72099025 on Build GRCh38
3957805 c.808C>G p.P270A Chr.16: 72098970-72098970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769585747 Chr.16: 72099007-72099007 on Build GRCh38
rs759882935 Chr.16: 72099165-72099165 on Build GRCh38
rs910317084 Chr.16: 72099124-72099124 on Build GRCh38
rs937863276 Chr.16: 72099196-72099196 on Build GRCh38
rs548505029 Chr.16: 72099016-72099016 on Build GRCh38
rs770139698 Chr.16: 72099168-72099168 on Build GRCh38
rs913780597 Chr.16: 72099139-72099140 on Build GRCh38
rs763619150 Chr.16: 72099073-72099073 on Build GRCh38
rs773314870 Chr.16: 72099026-72099026 on Build GRCh38
rs770617102 Chr.16: 72099161-72099161 on Build GRCh38
rs751278766 Chr.16: 72099081-72099081 on Build GRCh38
rs750236128 Chr.16: 72099090-72099090 on Build GRCh38
rs372876117 Chr.16: 72099153-72099153 on Build GRCh38
rs781084397 Chr.16: 72098997-72098997 on Build GRCh38
rs756947274 Chr.16: 72099087-72099087 on Build GRCh38
rs200290503 Chr.16: 72099088-72099088 on Build GRCh38
rs370793135 Chr.16: 72099043-72099043 on Build GRCh38
rs570497524 Chr.16: 72099146-72099146 on Build GRCh38
rs760079095 Chr.16: 72099030-72099030 on Build GRCh38
rs978181684 Chr.16: 72099119-72099119 on Build GRCh38
rs989119629 Chr.16: 72099136-72099136 on Build GRCh38
rs774553971 Chr.16: 72099008-72099008 on Build GRCh38
rs191078136 Chr.16: 72099000-72099000 on Build GRCh38
rs374300237 Chr.16: 72099095-72099095 on Build GRCh38
rs1046331390 Chr.16: 72099172-72099172 on Build GRCh38
rs1054171313 Chr.16: 72099151-72099151 on Build GRCh38
rs763560423 Chr.16: 72099209-72099209 on Build GRCh38
rs757149271 Chr.16: 72098996-72098996 on Build GRCh38
rs763467890 Chr.16: 72099187-72099187 on Build GRCh38
rs368516031 Chr.16: 72099210-72099210 on Build GRCh38
rs1044238899 Chr.16: 72099163-72099163 on Build GRCh38
rs377243883 Chr.16: 72099006-72099006 on Build GRCh38
rs748989584 Chr.16: 72099094-72099094 on Build GRCh38
rs377169556 Chr.16: 72099154-72099154 on Build GRCh38
rs143671341 Chr.16: 72099028-72099028 on Build GRCh38
rs775866943 Chr.16: 72099174-72099174 on Build GRCh38
rs954873751 Chr.16: 72099101-72099101 on Build GRCh38
rs924015733 Chr.16: 72099129-72099129 on Build GRCh38
rs146534594 Chr.16: 72099135-72099135 on Build GRCh38
rs556545076 Chr.16: 72099156-72099156 on Build GRCh38
rs148934675 Chr.16: 72099025-72099025 on Build GRCh38
rs760850084 Chr.16: 72099027-72099027 on Build GRCh38
rs866066908 Chr.16: 72099144-72099144 on Build GRCh38
rs574788791 Chr.16: 72099159-72099159 on Build GRCh38
rs776738831 Chr.16: 72099057-72099057 on Build GRCh38

View Full Product Details