Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56566507-56566780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC66
Entrez Gene ID
285331
Gene Name
coiled-coil domain containing 66
Gene Aliases
-
UniGene
Hs.476399
Gene Chromosome Location
Chr.3: 56557125-56621837 on Build GRCh38
Transcripts
NM_001012506.4 NM_001141947.1 NR_024460.1 XM_005265081.3 XM_005265082.3 XM_005265083.3 XM_005265084.3 XM_011533614.2 XM_011533615.2 XM_011533616.2 XM_011533617.2 XM_011533619.2 XM_017006229.1 XM_017006230.1 XM_017006231.1 XM_017006232.1 XM_017006233.1 XM_017006234.1 XM_017006235.1 XM_017006236.1 XM_017006237.1 XM_017006238.1 XM_017006239.1 XM_017006240.1 XM_017006241.1 XM_017006242.1 XM_017006243.1 XM_017006244.1 XM_017006245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5153858 c.612_613insA p.T207fs*2 Chr.3: 56566661-56566662 on Build GRCh38
6622357 c.697_698insA p.I235fs*6 Chr.3: 56566746-56566747 on Build GRCh38
4119450 c.637G>A p.V213I Chr.3: 56566686-56566686 on Build GRCh38
4337508 c.613delA p.T207fs*15 Chr.3: 56566662-56566662 on Build GRCh38
2781470 c.613delA p.T207fs*15 Chr.3: 56566662-56566662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370520912 Chr.3: 56566688-56566688 on Build GRCh38
rs199632321 Chr.3: 56566628-56566628 on Build GRCh38
rs1057190408 Chr.3: 56566716-56566716 on Build GRCh38
rs200227044 Chr.3: 56566648-56566648 on Build GRCh38
rs771507516 Chr.3: 56566647-56566647 on Build GRCh38
rs779130645 Chr.3: 56566598-56566598 on Build GRCh38
rs148949812 Chr.3: 56566662-56566662 on Build GRCh38
rs1011538597 Chr.3: 56566686-56566686 on Build GRCh38
rs771615087 Chr.3: 56566665-56566665 on Build GRCh38
rs749892170 Chr.3: 56566544-56566544 on Build GRCh38
rs746307461 Chr.3: 56566738-56566738 on Build GRCh38
rs760076568 Chr.3: 56566682-56566682 on Build GRCh38
rs757234007 Chr.3: 56566723-56566723 on Build GRCh38
rs760726995 Chr.3: 56566550-56566550 on Build GRCh38
rs148024341 Chr.3: 56566663-56566663 on Build GRCh38
rs749912342 Chr.3: 56566604-56566604 on Build GRCh38
rs374227528 Chr.3: 56566615-56566615 on Build GRCh38
rs770416445 Chr.3: 56566753-56566753 on Build GRCh38
rs78488477 Chr.3: 56566580-56566580 on Build GRCh38
rs1042342779 Chr.3: 56566701-56566701 on Build GRCh38
rs916784063 Chr.3: 56566589-56566589 on Build GRCh38
rs764946511 Chr.3: 56566702-56566702 on Build GRCh38
rs201070977 Chr.3: 56566707-56566707 on Build GRCh38
rs371765887 Chr.3: 56566714-56566714 on Build GRCh38
rs777975294 Chr.3: 56566621-56566621 on Build GRCh38
rs759341415 Chr.3: 56566693-56566693 on Build GRCh38
rs763480910 Chr.3: 56566566-56566566 on Build GRCh38
rs200541604 Chr.3: 56566717-56566717 on Build GRCh38
rs781387291 Chr.3: 56566739-56566740 on Build GRCh38
rs746254467 Chr.3: 56566649-56566649 on Build GRCh38
rs868747716 Chr.3: 56566560-56566560 on Build GRCh38
rs766373452 Chr.3: 56566683-56566683 on Build GRCh38
rs781727919 Chr.3: 56566730-56566730 on Build GRCh38
rs963991162 Chr.3: 56566565-56566565 on Build GRCh38
rs368224350 Chr.3: 56566681-56566681 on Build GRCh38
rs935360175 Chr.3: 56566658-56566658 on Build GRCh38
rs900342466 Chr.3: 56566705-56566705 on Build GRCh38
rs752860341 Chr.3: 56566611-56566611 on Build GRCh38
rs754730243 Chr.3: 56566592-56566592 on Build GRCh38

View Full Product Details