Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132196996-132197221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYSL4
Entrez Gene ID
10570
Gene Name
dihydropyrimidinase like 4
Gene Aliases
CRMP3, DRP-4, ULIP4
UniGene
Hs.100058
Gene Chromosome Location
Chr.10: 132184983-132205776 on Build GRCh38
Transcripts
NM_006426.2 XM_005252658.3 XM_011539125.2 XM_017015482.1 XM_017015483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1638483 c.563T>A p.I188N Chr.10: 132197043-132197043 on Build GRCh38
4012759 c.578C>T p.A193V Chr.10: 132197058-132197058 on Build GRCh38
5831428 c.553delT p.F185fs*71 Chr.10: 132197033-132197033 on Build GRCh38
1703256 c.547G>A p.E183K Chr.10: 132197027-132197027 on Build GRCh38
5734159 c.604G>A p.G202R Chr.10: 132197084-132197084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571990698 Chr.10: 132197123-132197123 on Build GRCh38
rs772052741 Chr.10: 132197065-132197065 on Build GRCh38
rs765584469 Chr.10: 132197086-132197086 on Build GRCh38
rs142662235 Chr.10: 132197195-132197195 on Build GRCh38
rs762940389 Chr.10: 132197115-132197137 on Build GRCh38
rs756820848 Chr.10: 132197122-132197122 on Build GRCh38
rs949378744 Chr.10: 132197202-132197202 on Build GRCh38
rs763682239 Chr.10: 132197135-132197151 on Build GRCh38
rs112243018 Chr.10: 132197109-132197109 on Build GRCh38
rs759388990 Chr.10: 132197052-132197053 on Build GRCh38
rs755952856 Chr.10: 132197045-132197045 on Build GRCh38
rs370569849 Chr.10: 132197013-132197013 on Build GRCh38
rs936746520 Chr.10: 132197124-132197125 on Build GRCh38
rs149176642 Chr.10: 132197083-132197083 on Build GRCh38
rs775889476 Chr.10: 132197087-132197087 on Build GRCh38
rs772394963 Chr.10: 132197187-132197187 on Build GRCh38
rs368882581 Chr.10: 132197085-132197085 on Build GRCh38
rs557478282 Chr.10: 132197103-132197118 on Build GRCh38
rs576572608 Chr.10: 132197074-132197074 on Build GRCh38
rs540651688 Chr.10: 132197135-132197135 on Build GRCh38
rs760228931 Chr.10: 132197084-132197084 on Build GRCh38
rs755894507 Chr.10: 132197139-132197139 on Build GRCh38
rs895139566 Chr.10: 132197151-132197151 on Build GRCh38
rs984482345 Chr.10: 132197039-132197039 on Build GRCh38
rs775569209 Chr.10: 132197103-132197141 on Build GRCh38
rs996653997 Chr.10: 132197106-132197106 on Build GRCh38
rs373050503 Chr.10: 132197186-132197186 on Build GRCh38
rs114629624 Chr.10: 132197107-132197107 on Build GRCh38
rs868588376 Chr.10: 132197064-132197064 on Build GRCh38
rs374798960 Chr.10: 132197026-132197026 on Build GRCh38
rs745699375 Chr.10: 132197041-132197041 on Build GRCh38
rs763451242 Chr.10: 132197093-132197093 on Build GRCh38
rs546417150 Chr.10: 132197046-132197046 on Build GRCh38
rs755004641 Chr.10: 132197146-132197146 on Build GRCh38
rs985075309 Chr.10: 132197130-132197130 on Build GRCh38
rs772648880 Chr.10: 132197032-132197032 on Build GRCh38
rs751170772 Chr.10: 132197120-132197120 on Build GRCh38
rs992357021 Chr.10: 132197179-132197179 on Build GRCh38
rs201152590 Chr.10: 132197144-132197144 on Build GRCh38
rs202122121 Chr.10: 132197051-132197051 on Build GRCh38
rs751260606 Chr.10: 132197023-132197023 on Build GRCh38

View Full Product Details