Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50312946-50313181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND6B
Entrez Gene ID
414918
Gene Name
DENN domain containing 6B
Gene Aliases
AFI1B, FAM116B
UniGene
Hs.530380
Gene Chromosome Location
Chr.22: 50309032-50327088 on Build GRCh38
Transcripts
NM_001001794.3 XM_005261914.3 XM_011530692.2 XM_011530693.2 XM_017028801.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4641182 c.1443G>T p.W481C Chr.22: 50313013-50313013 on Build GRCh38
6432570 c.1423T>C p.C475R Chr.22: 50313033-50313033 on Build GRCh38
7286876 c.1391G>A p.R464H Chr.22: 50313065-50313065 on Build GRCh38
1417088 c.1356C>G p.P452P Chr.22: 50313100-50313100 on Build GRCh38
4682408 c.1357delC p.Q453fs*55 Chr.22: 50313099-50313099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746588704 Chr.22: 50313065-50313065 on Build GRCh38
rs367677554 Chr.22: 50313145-50313145 on Build GRCh38
rs377653609 Chr.22: 50313066-50313066 on Build GRCh38
rs759816229 Chr.22: 50313005-50313005 on Build GRCh38
rs763517035 Chr.22: 50313116-50313116 on Build GRCh38
rs918732821 Chr.22: 50313043-50313043 on Build GRCh38
rs758090104 Chr.22: 50313062-50313062 on Build GRCh38
rs757154750 Chr.22: 50313041-50313041 on Build GRCh38
rs752840146 Chr.22: 50313027-50313027 on Build GRCh38
rs763173629 Chr.22: 50312994-50312994 on Build GRCh38
rs764192210 Chr.22: 50313149-50313149 on Build GRCh38
rs575835619 Chr.22: 50313164-50313164 on Build GRCh38
rs200021797 Chr.22: 50313134-50313134 on Build GRCh38
rs760696782 Chr.22: 50313148-50313148 on Build GRCh38
rs771141090 Chr.22: 50313127-50313127 on Build GRCh38
rs67248662 Chr.22: 50313100-50313100 on Build GRCh38
rs767569849 Chr.22: 50313019-50313019 on Build GRCh38
rs772127145 Chr.22: 50313099-50313099 on Build GRCh38
rs1027286215 Chr.22: 50313070-50313070 on Build GRCh38
rs542930160 Chr.22: 50313144-50313144 on Build GRCh38
rs1012663191 Chr.22: 50313098-50313099 on Build GRCh38
rs200103246 Chr.22: 50313099-50313099 on Build GRCh38
rs535004346 Chr.22: 50313112-50313112 on Build GRCh38
rs766794580 Chr.22: 50312995-50312995 on Build GRCh38
rs958454184 Chr.22: 50313080-50313080 on Build GRCh38
rs750397725 Chr.22: 50313060-50313060 on Build GRCh38
rs200746706 Chr.22: 50313129-50313129 on Build GRCh38
rs867591210 Chr.22: 50313021-50313021 on Build GRCh38
rs773812920 Chr.22: 50313103-50313103 on Build GRCh38
rs1028824105 Chr.22: 50313157-50313157 on Build GRCh38
rs201782165 Chr.22: 50313057-50313057 on Build GRCh38
rs769447051 Chr.22: 50313101-50313101 on Build GRCh38
rs562424306 Chr.22: 50313004-50313004 on Build GRCh38
rs151293382 Chr.22: 50313119-50313119 on Build GRCh38
rs1010298982 Chr.22: 50313122-50313122 on Build GRCh38
rs779905094 Chr.22: 50313064-50313064 on Build GRCh38
rs751663063 Chr.22: 50313001-50313001 on Build GRCh38

View Full Product Details