Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5914710-5914979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPS
Entrez Gene ID
828
Gene Name
calcyphosine
Gene Aliases
CAPS1
UniGene
Hs.584744
Gene Chromosome Location
Chr.19: 5913644-5916211 on Build GRCh38
Transcripts
NM_004058.4 NM_080590.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4850216 c.250C>T p.R84W Chr.19: 5914729-5914729 on Build GRCh38
7205317 c.263C>T p.P88L Chr.19: 5914941-5914941 on Build GRCh38
5973227 c.281G>A p.R94Q Chr.19: 5914959-5914959 on Build GRCh38
1002800 c.262-1G>T p.? Chr.19: 5914939-5914939 on Build GRCh38
6866473 c.243G>A p.E81E Chr.19: 5914722-5914722 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753951369 Chr.19: 5914812-5914812 on Build GRCh38
rs957637819 Chr.19: 5914807-5914807 on Build GRCh38
rs375700493 Chr.19: 5914959-5914959 on Build GRCh38
rs779696493 Chr.19: 5914951-5914951 on Build GRCh38
rs972745187 Chr.19: 5914938-5914938 on Build GRCh38
rs762476482 Chr.19: 5914787-5914787 on Build GRCh38
rs755044434 Chr.19: 5914757-5914757 on Build GRCh38
rs200346045 Chr.19: 5914785-5914785 on Build GRCh38
rs763056868 Chr.19: 5914898-5914898 on Build GRCh38
rs551938283 Chr.19: 5914799-5914799 on Build GRCh38
rs758702919 Chr.19: 5914748-5914748 on Build GRCh38
rs749240604 Chr.19: 5914765-5914765 on Build GRCh38
rs943754675 Chr.19: 5914873-5914873 on Build GRCh38
rs767592729 Chr.19: 5914915-5914915 on Build GRCh38
rs746645973 Chr.19: 5914955-5914955 on Build GRCh38
rs771793574 Chr.19: 5914776-5914776 on Build GRCh38
rs779977136 Chr.19: 5914750-5914750 on Build GRCh38
rs778058706 Chr.19: 5914738-5914738 on Build GRCh38
rs568391229 Chr.19: 5914931-5914931 on Build GRCh38
rs1023120001 Chr.19: 5914867-5914867 on Build GRCh38
rs913675251 Chr.19: 5914860-5914860 on Build GRCh38
rs755031185 Chr.19: 5914914-5914914 on Build GRCh38
rs61741511 Chr.19: 5914940-5914940 on Build GRCh38
rs79897803 Chr.19: 5914779-5914779 on Build GRCh38
rs756427610 Chr.19: 5914730-5914730 on Build GRCh38
rs181863983 Chr.19: 5914927-5914927 on Build GRCh38
rs56300912 Chr.19: 5914954-5914954 on Build GRCh38
rs372519827 Chr.19: 5914947-5914947 on Build GRCh38
rs745497707 Chr.19: 5914773-5914773 on Build GRCh38
rs750667426 Chr.19: 5914744-5914744 on Build GRCh38
rs777210281 Chr.19: 5914786-5914786 on Build GRCh38
rs766583938 Chr.19: 5914901-5914901 on Build GRCh38
rs777639752 Chr.19: 5914761-5914761 on Build GRCh38
rs957727512 Chr.19: 5914829-5914829 on Build GRCh38
rs977336644 Chr.19: 5914883-5914883 on Build GRCh38
rs376989134 Chr.19: 5914766-5914766 on Build GRCh38
rs773759817 Chr.19: 5914897-5914897 on Build GRCh38
rs778644633 Chr.19: 5914941-5914941 on Build GRCh38
rs373763376 Chr.19: 5914895-5914895 on Build GRCh38
rs568856885 Chr.19: 5914874-5914874 on Build GRCh38
rs546796912 Chr.19: 5914767-5914767 on Build GRCh38
rs921417121 Chr.19: 5914884-5914884 on Build GRCh38
rs749130772 Chr.19: 5914813-5914813 on Build GRCh38
rs868055405 Chr.19: 5914806-5914806 on Build GRCh38
rs939997811 Chr.19: 5914937-5914937 on Build GRCh38
rs987805486 Chr.19: 5914847-5914847 on Build GRCh38
rs376923370 Chr.19: 5914907-5914907 on Build GRCh38
rs138912753 Chr.19: 5914733-5914733 on Build GRCh38
rs754414584 Chr.19: 5914739-5914739 on Build GRCh38
rs768276521 Chr.19: 5914958-5914958 on Build GRCh38
rs758933554 Chr.19: 5914940-5914942 on Build GRCh38
rs11437855 Chr.19: 5914812-5914813 on Build GRCh38
rs765700900 Chr.19: 5914743-5914743 on Build GRCh38
rs760179767 Chr.19: 5914780-5914780 on Build GRCh38
rs764895542 Chr.19: 5914941-5914955 on Build GRCh38
rs776247074 Chr.19: 5914755-5914756 on Build GRCh38
rs981582949 Chr.19: 5914925-5914925 on Build GRCh38

View Full Product Details