Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16521606-16521831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHERP
Entrez Gene ID
10523
Gene Name
calcium homeostasis endoplasmic reticulum protein
Gene Aliases
DAN16, SCAF6, SRA1
UniGene
Hs.740364
Gene Chromosome Location
Chr.19: 16517889-16542452 on Build GRCh38
Transcripts
NM_006387.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1391275 c.1996T>C p.Y666H Chr.19: 16521639-16521639 on Build GRCh38
3530034 c.2028C>T p.L676L Chr.19: 16521607-16521607 on Build GRCh38
7200868 c.1981C>T p.L661L Chr.19: 16521654-16521654 on Build GRCh38
5789230 c.1981-9C>T p.? Chr.19: 16521663-16521663 on Build GRCh38
4075170 c.2002C>T p.P668S Chr.19: 16521633-16521633 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371699071 Chr.19: 16521643-16521643 on Build GRCh38
rs375715226 Chr.19: 16521659-16521659 on Build GRCh38
rs959462239 Chr.19: 16521808-16521808 on Build GRCh38
rs755582064 Chr.19: 16521625-16521625 on Build GRCh38
rs747075242 Chr.19: 16521705-16521705 on Build GRCh38
rs930155536 Chr.19: 16521795-16521795 on Build GRCh38
rs1020909433 Chr.19: 16521693-16521693 on Build GRCh38
rs375137581 Chr.19: 16521695-16521695 on Build GRCh38
rs530212877 Chr.19: 16521741-16521741 on Build GRCh38
rs770800905 Chr.19: 16521663-16521663 on Build GRCh38
rs370084080 Chr.19: 16521679-16521679 on Build GRCh38
rs769269464 Chr.19: 16521788-16521788 on Build GRCh38
rs754849942 Chr.19: 16521668-16521668 on Build GRCh38
rs984261909 Chr.19: 16521810-16521811 on Build GRCh38
rs910249087 Chr.19: 16521742-16521742 on Build GRCh38
rs549931677 Chr.19: 16521798-16521798 on Build GRCh38
rs139498762 Chr.19: 16521686-16521686 on Build GRCh38
rs775120092 Chr.19: 16521789-16521789 on Build GRCh38
rs527500961 Chr.19: 16521680-16521680 on Build GRCh38
rs776953905 Chr.19: 16521739-16521739 on Build GRCh38
rs986183604 Chr.19: 16521725-16521725 on Build GRCh38
rs759507094 Chr.19: 16521712-16521712 on Build GRCh38
rs964235491 Chr.19: 16521769-16521769 on Build GRCh38

View Full Product Details