Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31222871-31223126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_006719768.3 XM_011534936.1 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3813709 c.135T>C p.S45S Chr.13: 31222966-31222966 on Build GRCh38
1366325 c.138T>C p.G46G Chr.13: 31222969-31222969 on Build GRCh38
6609259 c.155A>G p.D52G Chr.13: 31222986-31222986 on Build GRCh38
946675 c.131A>C p.K44T Chr.13: 31222962-31222962 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs139243956 Chr.13: 31222897-31222897 on Build GRCh38
rs946759687 Chr.13: 31223031-31223031 on Build GRCh38
rs757186642 Chr.13: 31222968-31222968 on Build GRCh38
rs923943520 Chr.13: 31223073-31223073 on Build GRCh38
rs762380786 Chr.13: 31222920-31222920 on Build GRCh38
rs748910111 Chr.13: 31223013-31223013 on Build GRCh38
rs753993963 Chr.13: 31222948-31222948 on Build GRCh38
rs749099138 Chr.13: 31223060-31223060 on Build GRCh38
rs143166785 Chr.13: 31222911-31222911 on Build GRCh38
rs768120110 Chr.13: 31223014-31223014 on Build GRCh38
rs1015026893 Chr.13: 31223074-31223074 on Build GRCh38
rs769668416 Chr.13: 31223003-31223003 on Build GRCh38
rs202023402 Chr.13: 31222918-31222918 on Build GRCh38
rs755028268 Chr.13: 31222952-31222952 on Build GRCh38
rs779035727 Chr.13: 31222958-31222958 on Build GRCh38
rs752091076 Chr.13: 31223002-31223003 on Build GRCh38
rs775229706 Chr.13: 31222902-31222902 on Build GRCh38
rs377596461 Chr.13: 31222929-31222929 on Build GRCh38
rs773883651 Chr.13: 31223025-31223025 on Build GRCh38
rs767815041 Chr.13: 31222926-31222926 on Build GRCh38
rs755710240 Chr.13: 31223003-31223003 on Build GRCh38
rs374075796 Chr.13: 31223035-31223035 on Build GRCh38
rs1004703145 Chr.13: 31223058-31223058 on Build GRCh38
rs34510625 Chr.13: 31222978-31222979 on Build GRCh38
rs781181291 Chr.13: 31222985-31222985 on Build GRCh38
rs745676761 Chr.13: 31222986-31222986 on Build GRCh38
rs370713248 Chr.13: 31222946-31222946 on Build GRCh38
rs777482806 Chr.13: 31223033-31223033 on Build GRCh38
rs554134863 Chr.13: 31222944-31222944 on Build GRCh38
rs775283192 Chr.13: 31223010-31223010 on Build GRCh38
rs768038721 Chr.13: 31223030-31223030 on Build GRCh38
rs548514731 Chr.13: 31223085-31223085 on Build GRCh38
rs572785989 Chr.13: 31222960-31222960 on Build GRCh38
rs763590380 Chr.13: 31222915-31222915 on Build GRCh38

View Full Product Details