Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 128748659-128748896 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALU
Entrez Gene ID
813
Gene Name
calumenin
Gene Aliases
-
UniGene
Hs.743262
Gene Chromosome Location
Chr.7: 128739292-128773423 on Build GRCh38
Transcripts
NM_001130674.2 NM_001199671.1 NM_001199672.1 NM_001199673.1 NM_001219.4 NR_074086.1 XM_017012659.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6487794 c.76G>A p.D26N Chr.7: 128748659-128748659 on Build GRCh38
2769007 c.208G>T p.D70Y Chr.7: 128748767-128748767 on Build GRCh38
6847873 c.110A>C p.K37T Chr.7: 128748693-128748693 on Build GRCh38
1447901 c.195A>C p.T65T Chr.7: 128748778-128748778 on Build GRCh38
6487793 c.100G>A p.D34N Chr.7: 128748659-128748659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762263910 Chr.7: 128748848-128748848 on Build GRCh38
rs766116302 Chr.7: 128748742-128748742 on Build GRCh38
rs763096429 Chr.7: 128748828-128748828 on Build GRCh38
rs778336574 Chr.7: 128748778-128748778 on Build GRCh38
rs757709196 Chr.7: 128748708-128748708 on Build GRCh38
rs142806198 Chr.7: 128748691-128748691 on Build GRCh38
rs776040372 Chr.7: 128748685-128748685 on Build GRCh38
rs746752086 Chr.7: 128748803-128748803 on Build GRCh38
rs753501278 Chr.7: 128748751-128748751 on Build GRCh38
rs768035019 Chr.7: 128748850-128748850 on Build GRCh38
rs141712958 Chr.7: 128748779-128748779 on Build GRCh38
rs200127888 Chr.7: 128748683-128748683 on Build GRCh38
rs758295234 Chr.7: 128748787-128748787 on Build GRCh38
rs770774637 Chr.7: 128748814-128748814 on Build GRCh38
rs769226451 Chr.7: 128748834-128748834 on Build GRCh38
rs775055735 Chr.7: 128748842-128748842 on Build GRCh38
rs369536541 Chr.7: 128748824-128748824 on Build GRCh38
rs146270934 Chr.7: 128748693-128748693 on Build GRCh38
rs900197208 Chr.7: 128748829-128748829 on Build GRCh38
rs754480898 Chr.7: 128748772-128748772 on Build GRCh38
rs777695082 Chr.7: 128748800-128748800 on Build GRCh38
rs552027037 Chr.7: 128748815-128748815 on Build GRCh38
rs746121507 Chr.7: 128748819-128748819 on Build GRCh38
rs138128344 Chr.7: 128748702-128748702 on Build GRCh38

View Full Product Details