Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 23495790-23496015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CST8
Entrez Gene ID
10047
Gene Name
cystatin 8
Gene Aliases
CRES, CTES5
UniGene
Hs.121602
Gene Chromosome Location
Chr.20: 23491077-23507124 on Build GRCh38
Transcripts
NM_001281730.1 NM_005492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707369 c.377T>G p.L126R Chr.20: 23495862-23495862 on Build GRCh38
6159604 c.381C>T p.P127P Chr.20: 23495866-23495866 on Build GRCh38
6871385 c.354C>T p.S118S Chr.20: 23495839-23495839 on Build GRCh38
6637000 c.346-1G>A p.? Chr.20: 23495830-23495830 on Build GRCh38
5973977 c.346A>G p.K116E Chr.20: 23495831-23495831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375674608 Chr.20: 23495890-23495890 on Build GRCh38
rs764168823 Chr.20: 23495914-23495914 on Build GRCh38
rs749175564 Chr.20: 23495929-23495929 on Build GRCh38
rs892652951 Chr.20: 23495980-23495980 on Build GRCh38
rs747197475 Chr.20: 23495840-23495840 on Build GRCh38
rs780017021 Chr.20: 23495927-23495927 on Build GRCh38
rs373305124 Chr.20: 23495822-23495822 on Build GRCh38
rs1043817058 Chr.20: 23495990-23495990 on Build GRCh38
rs377479607 Chr.20: 23495855-23495855 on Build GRCh38
rs997261763 Chr.20: 23495970-23495970 on Build GRCh38
rs946868033 Chr.20: 23495988-23495988 on Build GRCh38
rs760346876 Chr.20: 23495950-23495950 on Build GRCh38
rs771727843 Chr.20: 23495948-23495948 on Build GRCh38
rs1030012324 Chr.20: 23495986-23495986 on Build GRCh38
rs556293524 Chr.20: 23495971-23495971 on Build GRCh38
rs772815329 Chr.20: 23495949-23495949 on Build GRCh38
rs750627921 Chr.20: 23495850-23495850 on Build GRCh38
rs750211203 Chr.20: 23495921-23495921 on Build GRCh38
rs755932425 Chr.20: 23495926-23495926 on Build GRCh38
rs769830855 Chr.20: 23495851-23495851 on Build GRCh38
rs781038519 Chr.20: 23495845-23495845 on Build GRCh38
rs1054633 Chr.20: 23495909-23495909 on Build GRCh38
rs372473753 Chr.20: 23495857-23495857 on Build GRCh38
rs547013395 Chr.20: 23495939-23495939 on Build GRCh38
rs761459406 Chr.20: 23495874-23495874 on Build GRCh38
rs777740748 Chr.20: 23495838-23495838 on Build GRCh38
rs752116617 Chr.20: 23495933-23495933 on Build GRCh38
rs753158690 Chr.20: 23495901-23495901 on Build GRCh38
rs368837269 Chr.20: 23495891-23495891 on Build GRCh38
rs372313950 Chr.20: 23495915-23495915 on Build GRCh38
rs765784244 Chr.20: 23495819-23495819 on Build GRCh38
rs984559285 Chr.20: 23495905-23495905 on Build GRCh38
rs1802329 Chr.20: 23495903-23495903 on Build GRCh38
rs140211912 Chr.20: 23495977-23495977 on Build GRCh38
rs376287858 Chr.20: 23495889-23495889 on Build GRCh38
rs781316844 Chr.20: 23495920-23495920 on Build GRCh38
rs773743932 Chr.20: 23495865-23495865 on Build GRCh38
rs200344286 Chr.20: 23495931-23495931 on Build GRCh38
rs202124724 Chr.20: 23495964-23495964 on Build GRCh38
rs559058456 Chr.20: 23495859-23495859 on Build GRCh38
rs201638851 Chr.20: 23495843-23495843 on Build GRCh38
rs1006496784 Chr.20: 23495917-23495917 on Build GRCh38
rs73901886 Chr.20: 23495965-23495965 on Build GRCh38
rs766846477 Chr.20: 23495902-23495902 on Build GRCh38
rs765624860 Chr.20: 23495897-23495897 on Build GRCh38
rs976147582 Chr.20: 23495960-23495960 on Build GRCh38

View Full Product Details