Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 64044787-64045012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERN1
Entrez Gene ID
2081
Gene Name
endoplasmic reticulum to nucleus signaling 1
Gene Aliases
IRE1, IRE1P, IRE1a, hIRE1p
UniGene
Hs.133982
Gene Chromosome Location
Chr.17: 64039142-64130819 on Build GRCh38
Transcripts
NM_001433.3 XM_017024347.1 XM_017024348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5573609 c.2654-1G>A p.? Chr.17: 64044928-64044928 on Build GRCh38
7235893 c.2654-10G>A p.? Chr.17: 64044937-64044937 on Build GRCh38
1236297 c.2714G>C p.R905T Chr.17: 64044867-64044867 on Build GRCh38
6213805 c.2703C>T p.L901L Chr.17: 64044878-64044878 on Build GRCh38
6993376 c.2667C>T p.F889F Chr.17: 64044914-64044914 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs114005448 Chr.17: 64044814-64044814 on Build GRCh38
rs374579165 Chr.17: 64044819-64044819 on Build GRCh38
rs371950088 Chr.17: 64044922-64044922 on Build GRCh38
rs756111954 Chr.17: 64044932-64044932 on Build GRCh38
rs761267281 Chr.17: 64044822-64044822 on Build GRCh38
rs750671350 Chr.17: 64044902-64044902 on Build GRCh38
rs753989451 Chr.17: 64044851-64044851 on Build GRCh38
rs761261557 Chr.17: 64044945-64044945 on Build GRCh38
rs765615060 Chr.17: 64044877-64044877 on Build GRCh38
rs748881223 Chr.17: 64044956-64044956 on Build GRCh38
rs777544229 Chr.17: 64044934-64044934 on Build GRCh38
rs368195187 Chr.17: 64044876-64044876 on Build GRCh38
rs764467817 Chr.17: 64044828-64044828 on Build GRCh38
rs991714289 Chr.17: 64044971-64044971 on Build GRCh38
rs758628467 Chr.17: 64044921-64044921 on Build GRCh38
rs552137526 Chr.17: 64044816-64044816 on Build GRCh38
rs747907366 Chr.17: 64044931-64044931 on Build GRCh38

View Full Product Details