Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12677154-12677425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHPS
Entrez Gene ID
1725
Gene Name
deoxyhypusine synthase
Gene Aliases
DHS, DS, MIG13
UniGene
Hs.79064
Gene Chromosome Location
Chr.19: 12672164-12681887 on Build GRCh38
Transcripts
NM_001206974.1 NM_001930.3 NM_013406.2 NR_038192.1 XM_011527770.1 XM_011527771.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982861 c.715G>T p.G239C Chr.19: 12677360-12677360 on Build GRCh38
438620 c.697A>C p.S233R Chr.19: 12677378-12677378 on Build GRCh38
6643294 c.696T>C p.F232F Chr.19: 12677379-12677379 on Build GRCh38
991400 c.824G>A p.C275Y Chr.19: 12677172-12677172 on Build GRCh38
4507461 c.746C>T p.S249F Chr.19: 12677329-12677329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370385403 Chr.19: 12677272-12677272 on Build GRCh38
rs376157066 Chr.19: 12677284-12677284 on Build GRCh38
rs764549648 Chr.19: 12677366-12677366 on Build GRCh38
rs750681585 Chr.19: 12677375-12677375 on Build GRCh38
rs761549753 Chr.19: 12677345-12677345 on Build GRCh38
rs778230502 Chr.19: 12677300-12677300 on Build GRCh38
rs189109985 Chr.19: 12677283-12677283 on Build GRCh38
rs756762989 Chr.19: 12677365-12677366 on Build GRCh38
rs575787787 Chr.19: 12677189-12677189 on Build GRCh38
rs758418180 Chr.19: 12677190-12677190 on Build GRCh38
rs753258751 Chr.19: 12677297-12677297 on Build GRCh38
rs764915726 Chr.19: 12677271-12677271 on Build GRCh38
rs1012142857 Chr.19: 12677387-12677387 on Build GRCh38
rs535857791 Chr.19: 12677360-12677360 on Build GRCh38
rs756511088 Chr.19: 12677298-12677298 on Build GRCh38
rs749528954 Chr.19: 12677224-12677224 on Build GRCh38
rs745985350 Chr.19: 12677228-12677228 on Build GRCh38
rs547740533 Chr.19: 12677372-12677372 on Build GRCh38
rs765716761 Chr.19: 12677266-12677268 on Build GRCh38
rs919971126 Chr.19: 12677344-12677344 on Build GRCh38
rs974776882 Chr.19: 12677252-12677252 on Build GRCh38
rs754205298 Chr.19: 12677309-12677309 on Build GRCh38
rs568560231 Chr.19: 12677230-12677230 on Build GRCh38
rs757666183 Chr.19: 12677314-12677314 on Build GRCh38
rs746075617 Chr.19: 12677317-12677317 on Build GRCh38
rs113571246 Chr.19: 12677342-12677342 on Build GRCh38
rs773694823 Chr.19: 12677361-12677361 on Build GRCh38
rs780759776 Chr.19: 12677385-12677386 on Build GRCh38
rs760044789 Chr.19: 12677183-12677183 on Build GRCh38
rs367826439 Chr.19: 12677388-12677388 on Build GRCh38
rs912160587 Chr.19: 12677214-12677214 on Build GRCh38
rs374500589 Chr.19: 12677373-12677373 on Build GRCh38
rs751094326 Chr.19: 12677334-12677336 on Build GRCh38
rs938956428 Chr.19: 12677370-12677370 on Build GRCh38
rs758656990 Chr.19: 12677385-12677385 on Build GRCh38
rs780219273 Chr.19: 12677328-12677328 on Build GRCh38
rs781692345 Chr.19: 12677193-12677193 on Build GRCh38
rs371375475 Chr.19: 12677262-12677262 on Build GRCh38
rs772294853 Chr.19: 12677326-12677326 on Build GRCh38
rs934680485 Chr.19: 12677178-12677178 on Build GRCh38
rs779347086 Chr.19: 12677316-12677316 on Build GRCh38
rs761265606 Chr.19: 12677365-12677365 on Build GRCh38
rs753137312 Chr.19: 12677216-12677216 on Build GRCh38
rs565337083 Chr.19: 12677223-12677223 on Build GRCh38
rs762534456 Chr.19: 12677356-12677356 on Build GRCh38
rs781054297 Chr.19: 12677394-12677394 on Build GRCh38
rs377182954 Chr.19: 12677268-12677268 on Build GRCh38
rs755051406 Chr.19: 12677391-12677391 on Build GRCh38
rs143117686 Chr.19: 12677355-12677355 on Build GRCh38
rs373000198 Chr.19: 12677227-12677227 on Build GRCh38
rs761309007 Chr.19: 12677187-12677187 on Build GRCh38
rs747003113 Chr.19: 12677337-12677337 on Build GRCh38
rs1028652737 Chr.19: 12677254-12677254 on Build GRCh38
rs776604750 Chr.19: 12677340-12677340 on Build GRCh38
rs181620939 Chr.19: 12677402-12677402 on Build GRCh38
rs775628435 Chr.19: 12677235-12677235 on Build GRCh38
rs550355399 Chr.19: 12677183-12677183 on Build GRCh38
rs763786266 Chr.19: 12677177-12677177 on Build GRCh38
rs756557667 Chr.19: 12677222-12677222 on Build GRCh38
rs776254007 Chr.19: 12677267-12677267 on Build GRCh38
rs75392871 Chr.19: 12677225-12677225 on Build GRCh38
rs754366240 Chr.19: 12677371-12677371 on Build GRCh38
rs757878296 Chr.19: 12677191-12677191 on Build GRCh38
rs371100778 Chr.19: 12677349-12677349 on Build GRCh38
rs764762126 Chr.19: 12677188-12677188 on Build GRCh38
rs748062148 Chr.19: 12677397-12677397 on Build GRCh38

View Full Product Details