Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30930023-30930254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001099784.2 NM_001282351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6661923 c.1002C>T p.S334S Chr.16: 30930225-30930225 on Build GRCh38
7334198 c.894G>A p.P298P Chr.16: 30930117-30930117 on Build GRCh38
6661919 c.926G>A p.R309H Chr.16: 30930149-30930149 on Build GRCh38
4514980 c.993C>T p.D331D Chr.16: 30930216-30930216 on Build GRCh38
4563674 c.984G>A p.S328S Chr.16: 30930207-30930207 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746760154 Chr.16: 30930233-30930233 on Build GRCh38
rs943809386 Chr.16: 30930158-30930158 on Build GRCh38
rs760075736 Chr.16: 30930149-30930149 on Build GRCh38
rs200509975 Chr.16: 30930111-30930111 on Build GRCh38
rs755482038 Chr.16: 30930070-30930070 on Build GRCh38
rs932317485 Chr.16: 30930225-30930225 on Build GRCh38
rs958686076 Chr.16: 30930061-30930061 on Build GRCh38
rs565184862 Chr.16: 30930122-30930122 on Build GRCh38
rs753046805 Chr.16: 30930168-30930168 on Build GRCh38
rs768420902 Chr.16: 30930234-30930234 on Build GRCh38
rs569746984 Chr.16: 30930207-30930207 on Build GRCh38
rs369314821 Chr.16: 30930123-30930123 on Build GRCh38
rs532569356 Chr.16: 30930126-30930126 on Build GRCh38
rs771156225 Chr.16: 30930117-30930117 on Build GRCh38
rs773286265 Chr.16: 30930213-30930214 on Build GRCh38
rs969911741 Chr.16: 30930065-30930065 on Build GRCh38
rs987106518 Chr.16: 30930157-30930157 on Build GRCh38
rs764360734 Chr.16: 30930194-30930194 on Build GRCh38
rs761346687 Chr.16: 30930058-30930058 on Build GRCh38
rs749339084 Chr.16: 30930116-30930116 on Build GRCh38
rs776454702 Chr.16: 30930236-30930236 on Build GRCh38
rs764777094 Chr.16: 30930147-30930147 on Build GRCh38
rs779858487 Chr.16: 30930220-30930220 on Build GRCh38
rs752087489 Chr.16: 30930067-30930067 on Build GRCh38
rs761506046 Chr.16: 30930141-30930141 on Build GRCh38
rs772664806 Chr.16: 30930148-30930148 on Build GRCh38
rs779042030 Chr.16: 30930213-30930213 on Build GRCh38
rs757460088 Chr.16: 30930209-30930209 on Build GRCh38
rs775339194 Chr.16: 30930128-30930128 on Build GRCh38
rs753061922 Chr.16: 30930084-30930084 on Build GRCh38
rs746817391 Chr.16: 30930129-30930129 on Build GRCh38
rs768259565 Chr.16: 30930132-30930132 on Build GRCh38
rs528788728 Chr.16: 30930059-30930059 on Build GRCh38
rs368537998 Chr.16: 30930046-30930046 on Build GRCh38
rs767923884 Chr.16: 30930159-30930159 on Build GRCh38
rs983034034 Chr.16: 30930121-30930121 on Build GRCh38
rs781616493 Chr.16: 30930083-30930083 on Build GRCh38
rs756348504 Chr.16: 30930110-30930110 on Build GRCh38
rs756575659 Chr.16: 30930193-30930193 on Build GRCh38
rs199802404 Chr.16: 30930135-30930135 on Build GRCh38
rs745989912 Chr.16: 30930219-30930219 on Build GRCh38
rs962468553 Chr.16: 30930108-30930108 on Build GRCh38

View Full Product Details