Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 40153432-40153699 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf4
Entrez Gene ID
56892
Gene Name
chromosome 8 open reading frame 4
Gene Aliases
TC-1, TC1
UniGene
Hs.591849
Gene Chromosome Location
Chr.8: 40153468-40155308 on Build GRCh38
Transcripts
NM_020130.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5932199 c.149G>A p.R50K Chr.8: 40153681-40153681 on Build GRCh38
5802847 c.149G>T p.R50M Chr.8: 40153681-40153681 on Build GRCh38
6189352 c.145G>A p.E49K Chr.8: 40153677-40153677 on Build GRCh38
3900134 c.152T>C p.L51P Chr.8: 40153684-40153684 on Build GRCh38
6013141 c.140C>T p.S47L Chr.8: 40153672-40153672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182358217 Chr.8: 40153484-40153484 on Build GRCh38
rs146758200 Chr.8: 40153627-40153627 on Build GRCh38
rs570222497 Chr.8: 40153638-40153638 on Build GRCh38
rs895047380 Chr.8: 40153567-40153567 on Build GRCh38
rs762024125 Chr.8: 40153613-40153613 on Build GRCh38
rs140361573 Chr.8: 40153631-40153631 on Build GRCh38
rs758408259 Chr.8: 40153546-40153546 on Build GRCh38
rs200396747 Chr.8: 40153515-40153515 on Build GRCh38
rs550162388 Chr.8: 40153589-40153589 on Build GRCh38
rs367776739 Chr.8: 40153617-40153617 on Build GRCh38
rs781484363 Chr.8: 40153665-40153665 on Build GRCh38
rs929523214 Chr.8: 40153460-40153460 on Build GRCh38
rs777075547 Chr.8: 40153610-40153610 on Build GRCh38
rs1012127429 Chr.8: 40153607-40153607 on Build GRCh38
rs772879844 Chr.8: 40153615-40153616 on Build GRCh38
rs931233400 Chr.8: 40153547-40153547 on Build GRCh38
rs369320336 Chr.8: 40153545-40153545 on Build GRCh38
rs749022444 Chr.8: 40153506-40153506 on Build GRCh38
rs11551219 Chr.8: 40153667-40153667 on Build GRCh38
rs769983492 Chr.8: 40153482-40153482 on Build GRCh38
rs6474226 Chr.8: 40153560-40153560 on Build GRCh38
rs762125414 Chr.8: 40153539-40153539 on Build GRCh38
rs771763209 Chr.8: 40153581-40153581 on Build GRCh38
rs200284520 Chr.8: 40153520-40153520 on Build GRCh38
rs765326334 Chr.8: 40153542-40153542 on Build GRCh38
rs370896349 Chr.8: 40153626-40153626 on Build GRCh38
rs761324185 Chr.8: 40153602-40153603 on Build GRCh38
rs747943481 Chr.8: 40153502-40153502 on Build GRCh38
rs781051504 Chr.8: 40153500-40153500 on Build GRCh38
rs773491577 Chr.8: 40153624-40153624 on Build GRCh38
rs757064401 Chr.8: 40153643-40153643 on Build GRCh38
rs755971691 Chr.8: 40153503-40153503 on Build GRCh38
rs773985483 Chr.8: 40153559-40153560 on Build GRCh38
rs777418368 Chr.8: 40153505-40153505 on Build GRCh38
rs773946763 Chr.8: 40153516-40153516 on Build GRCh38
rs779742891 Chr.8: 40153648-40153648 on Build GRCh38
rs752690598 Chr.8: 40153558-40153558 on Build GRCh38
rs770062071 Chr.8: 40153614-40153614 on Build GRCh38
rs745509044 Chr.8: 40153576-40153576 on Build GRCh38
rs766443410 Chr.8: 40153555-40153555 on Build GRCh38
rs777667508 Chr.8: 40153563-40153563 on Build GRCh38
rs753683431 Chr.8: 40153640-40153640 on Build GRCh38
rs1024069965 Chr.8: 40153637-40153637 on Build GRCh38
rs1129246 Chr.8: 40153570-40153570 on Build GRCh38
rs750145904 Chr.8: 40153645-40153645 on Build GRCh38
rs754929477 Chr.8: 40153494-40153494 on Build GRCh38
rs779684082 Chr.8: 40153582-40153582 on Build GRCh38
rs758089376 Chr.8: 40153647-40153647 on Build GRCh38
rs35273913 Chr.8: 40153657-40153657 on Build GRCh38
rs776829094 Chr.8: 40153538-40153538 on Build GRCh38
rs10353 Chr.8: 40153668-40153668 on Build GRCh38
rs776203347 Chr.8: 40153529-40153529 on Build GRCh38
rs150354849 Chr.8: 40153670-40153670 on Build GRCh38
rs772708294 Chr.8: 40153528-40153528 on Build GRCh38
rs769002993 Chr.8: 40153601-40153601 on Build GRCh38
rs749000656 Chr.8: 40153569-40153569 on Build GRCh38
rs778375012 Chr.8: 40153574-40153574 on Build GRCh38
rs539318737 Chr.8: 40153661-40153661 on Build GRCh38
rs750429059 Chr.8: 40153489-40153489 on Build GRCh38
rs761246190 Chr.8: 40153531-40153531 on Build GRCh38
rs764565156 Chr.8: 40153532-40153532 on Build GRCh38

View Full Product Details