Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 15816748-15817009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188A
Entrez Gene ID
80013
Gene Name
family with sequence similarity 188 member A
Gene Aliases
C10orf97, CARP, DERP5, MST126, MSTP126, my042
UniGene
Hs.158870
Gene Chromosome Location
Chr.10: 15778169-15860533 on Build GRCh38
Transcripts
NM_001318330.1 NM_024948.3 XM_005252600.2 XM_005252602.2 XM_006717508.2 XM_011519690.2 XM_011519691.2 XM_011519693.1 XM_011519694.1 XM_017016671.1 XM_017016672.1 XM_017016673.1 XM_017016674.1 XM_017016675.1 XM_017016676.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2131467 c.435delT p.F145fs*7 Chr.10: 15816841-15816841 on Build GRCh38
6247388 c.390C>A p.F130L Chr.10: 15816886-15816886 on Build GRCh38
4572804 c.871T>A p.F291I Chr.10: 15816846-15816846 on Build GRCh38
1638502 c.409A>G p.S137G Chr.10: 15816867-15816867 on Build GRCh38
6247387 c.831C>A p.F277L Chr.10: 15816886-15816886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760457587 Chr.10: 15816850-15816850 on Build GRCh38
rs115072884 Chr.10: 15816945-15816945 on Build GRCh38
rs996828268 Chr.10: 15816977-15816977 on Build GRCh38
rs771847937 Chr.10: 15816847-15816847 on Build GRCh38
rs925603199 Chr.10: 15816835-15816835 on Build GRCh38
rs766046506 Chr.10: 15816882-15816883 on Build GRCh38
rs773192645 Chr.10: 15816849-15816849 on Build GRCh38
rs1047204900 Chr.10: 15816941-15816941 on Build GRCh38
rs773102597 Chr.10: 15816936-15816936 on Build GRCh38
rs768696203 Chr.10: 15816918-15816918 on Build GRCh38
rs759060381 Chr.10: 15816882-15816882 on Build GRCh38
rs941438025 Chr.10: 15816947-15816947 on Build GRCh38
rs757935235 Chr.10: 15816894-15816894 on Build GRCh38
rs201669904 Chr.10: 15816887-15816887 on Build GRCh38
rs954628302 Chr.10: 15816782-15816782 on Build GRCh38
rs770876592 Chr.10: 15816951-15816951 on Build GRCh38
rs759133323 Chr.10: 15816946-15816949 on Build GRCh38
rs760653499 Chr.10: 15816946-15816946 on Build GRCh38
rs552709677 Chr.10: 15816980-15816980 on Build GRCh38
rs950313658 Chr.10: 15816898-15816898 on Build GRCh38
rs370002522 Chr.10: 15816903-15816903 on Build GRCh38
rs114035593 Chr.10: 15816960-15816960 on Build GRCh38
rs760976202 Chr.10: 15816808-15816809 on Build GRCh38
rs991116269 Chr.10: 15816857-15816857 on Build GRCh38
rs776366672 Chr.10: 15816958-15816958 on Build GRCh38
rs182755381 Chr.10: 15816790-15816790 on Build GRCh38
rs957532083 Chr.10: 15816779-15816779 on Build GRCh38
rs779208802 Chr.10: 15816922-15816922 on Build GRCh38
rs967515853 Chr.10: 15816808-15816808 on Build GRCh38
rs761960087 Chr.10: 15816819-15816819 on Build GRCh38
rs756528150 Chr.10: 15816908-15816908 on Build GRCh38
rs773585371 Chr.10: 15816809-15816809 on Build GRCh38
rs148141305 Chr.10: 15816915-15816915 on Build GRCh38
rs374105430 Chr.10: 15816925-15816925 on Build GRCh38
rs766202097 Chr.10: 15816866-15816866 on Build GRCh38
rs201380851 Chr.10: 15816798-15816798 on Build GRCh38
rs753464168 Chr.10: 15816877-15816877 on Build GRCh38
rs186662806 Chr.10: 15816978-15816978 on Build GRCh38
rs80206007 Chr.10: 15816817-15816817 on Build GRCh38
rs371814494 Chr.10: 15816793-15816793 on Build GRCh38
rs775828732 Chr.10: 15816796-15816796 on Build GRCh38
rs772259645 Chr.10: 15816926-15816926 on Build GRCh38
rs780520362 Chr.10: 15816913-15816913 on Build GRCh38
rs1020159917 Chr.10: 15816802-15816802 on Build GRCh38
rs201974094 Chr.10: 15816809-15816809 on Build GRCh38
rs751011612 Chr.10: 15816904-15816904 on Build GRCh38
rs768949057 Chr.10: 15816816-15816816 on Build GRCh38
rs776231187 Chr.10: 15816942-15816945 on Build GRCh38
rs890365276 Chr.10: 15816980-15816980 on Build GRCh38
rs749695338 Chr.10: 15816916-15816916 on Build GRCh38

View Full Product Details