Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111682051-111682324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
UniGene
Hs.443061
Gene Chromosome Location
Chr.X: 111680741-111764164 on Build GRCh38
Transcripts
NM_001039210.4 NM_001099922.2 NM_001168385.2 NM_001257230.1 NM_001257231.1 NM_001257234.1 NM_001257235.2 NM_001257237.1 NM_001257239.2 NM_001257240.2 NM_001257241.2 NM_001324290.1 NM_001324291.1 NM_001324292.1 NM_001324293.1 NM_001324294.1 NM_018466.5 NR_033125.2 XM_006724693.2 XM_006724697.3 XM_006724698.3 XM_011531028.2 XM_011531030.1 XM_011531031.1 XM_011531032.1 XM_011531033.2 XM_017029846.1 XM_017029847.1 XM_017029848.1 XM_017029849.1 XM_017029850.1 XM_017029851.1

SNP Detail

SNP ID Chromosome Location
rs777630603 Chr.X: 111682089-111682089 on Build GRCh38
rs11545580 Chr.X: 111682163-111682163 on Build GRCh38
rs768124065 Chr.X: 111682109-111682109 on Build GRCh38
rs762337561 Chr.X: 111682142-111682142 on Build GRCh38
rs765437651 Chr.X: 111682222-111682222 on Build GRCh38
rs141162495 Chr.X: 111682105-111682105 on Build GRCh38
rs147897682 Chr.X: 111682200-111682200 on Build GRCh38
rs752909568 Chr.X: 111682226-111682226 on Build GRCh38
rs966548065 Chr.X: 111682129-111682129 on Build GRCh38
rs146925326 Chr.X: 111682233-111682233 on Build GRCh38
rs369213082 Chr.X: 111682103-111682103 on Build GRCh38
rs774810032 Chr.X: 111682107-111682107 on Build GRCh38
rs199717765 Chr.X: 111682122-111682122 on Build GRCh38
rs769838471 Chr.X: 111682158-111682158 on Build GRCh38
rs149808406 Chr.X: 111682154-111682154 on Build GRCh38
rs1009722243 Chr.X: 111682139-111682139 on Build GRCh38
rs766512298 Chr.X: 111682189-111682189 on Build GRCh38
rs757032019 Chr.X: 111682095-111682095 on Build GRCh38
rs745826764 Chr.X: 111682257-111682257 on Build GRCh38
rs372304203 Chr.X: 111682205-111682205 on Build GRCh38
rs781170536 Chr.X: 111682096-111682096 on Build GRCh38
rs746749937 Chr.X: 111682090-111682090 on Build GRCh38
rs780942708 Chr.X: 111682238-111682238 on Build GRCh38
rs755115889 Chr.X: 111682085-111682085 on Build GRCh38
rs762379771 Chr.X: 111682098-111682099 on Build GRCh38
rs755960706 Chr.X: 111682261-111682261 on Build GRCh38
rs1057519175 Chr.X: 111682178-111682178 on Build GRCh38

View Full Product Details