Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1312089-1312352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3L
Entrez Gene ID
54973
Gene Name
cleavage and polyadenylation specific factor 3-like
Gene Aliases
CPSF73L, INT11, INTS11, RC-68, RC68
UniGene
Hs.6449
Gene Chromosome Location
Chr.1: 1311585-1324687 on Build GRCh38
Transcripts
NM_001256456.1 NM_001256460.1 NM_001256462.1 NM_001256463.1 NM_017871.5 XM_011541647.1 XM_011541648.1 XM_011541650.1 XM_017001557.1 XM_017001558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7288429 c.1588G>A p.V530I Chr.1: 1312245-1312245 on Build GRCh38
4880150 c.1640C>A p.P547Q Chr.1: 1312115-1312115 on Build GRCh38
6379373 c.1616A>C p.H539P Chr.1: 1312235-1312235 on Build GRCh38
6379372 c.1598A>C p.H533P Chr.1: 1312235-1312235 on Build GRCh38
7244371 c.1641A>G p.P547P Chr.1: 1312114-1312114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781305263 Chr.1: 1312129-1312129 on Build GRCh38
rs747804290 Chr.1: 1312280-1312280 on Build GRCh38
rs897239384 Chr.1: 1312237-1312237 on Build GRCh38
rs534327625 Chr.1: 1312224-1312224 on Build GRCh38
rs771585917 Chr.1: 1312223-1312223 on Build GRCh38
rs779962156 Chr.1: 1312210-1312210 on Build GRCh38
rs148516711 Chr.1: 1312111-1312111 on Build GRCh38
rs868284619 Chr.1: 1312244-1312244 on Build GRCh38
rs374446682 Chr.1: 1312255-1312255 on Build GRCh38
rs746897636 Chr.1: 1312116-1312116 on Build GRCh38
rs1004770917 Chr.1: 1312206-1312207 on Build GRCh38
rs568445177 Chr.1: 1312208-1312208 on Build GRCh38
rs764250347 Chr.1: 1312314-1312314 on Build GRCh38
rs374869753 Chr.1: 1312170-1312170 on Build GRCh38
rs759845797 Chr.1: 1312247-1312247 on Build GRCh38
rs751486747 Chr.1: 1312126-1312127 on Build GRCh38
rs767846766 Chr.1: 1312109-1312109 on Build GRCh38
rs757170283 Chr.1: 1312180-1312181 on Build GRCh38
rs776997048 Chr.1: 1312201-1312202 on Build GRCh38
rs748874135 Chr.1: 1312214-1312214 on Build GRCh38
rs777021763 Chr.1: 1312292-1312292 on Build GRCh38
rs764339825 Chr.1: 1312256-1312256 on Build GRCh38
rs12103 Chr.1: 1312114-1312114 on Build GRCh38
rs755900289 Chr.1: 1312199-1312199 on Build GRCh38
rs750952016 Chr.1: 1312167-1312167 on Build GRCh38
rs756503541 Chr.1: 1312110-1312110 on Build GRCh38
rs767558936 Chr.1: 1312259-1312259 on Build GRCh38
rs886336312 Chr.1: 1312201-1312201 on Build GRCh38
rs759883842 Chr.1: 1312293-1312293 on Build GRCh38
rs745607228 Chr.1: 1312131-1312131 on Build GRCh38
rs945229819 Chr.1: 1312135-1312135 on Build GRCh38
rs779686729 Chr.1: 1312264-1312264 on Build GRCh38
rs560124260 Chr.1: 1312108-1312108 on Build GRCh38
rs954645689 Chr.1: 1312217-1312217 on Build GRCh38
rs553875387 Chr.1: 1312243-1312243 on Build GRCh38
rs778824691 Chr.1: 1312196-1312196 on Build GRCh38
rs745807682 Chr.1: 1312209-1312209 on Build GRCh38
rs770228406 Chr.1: 1312203-1312204 on Build GRCh38
rs773963445 Chr.1: 1312156-1312156 on Build GRCh38
rs949448745 Chr.1: 1312308-1312308 on Build GRCh38
rs752778156 Chr.1: 1312204-1312204 on Build GRCh38
rs764278418 Chr.1: 1312263-1312263 on Build GRCh38
rs768042214 Chr.1: 1312151-1312151 on Build GRCh38
rs769722262 Chr.1: 1312144-1312144 on Build GRCh38
rs372959391 Chr.1: 1312149-1312149 on Build GRCh38
rs977911026 Chr.1: 1312139-1312139 on Build GRCh38
rs763327656 Chr.1: 1312312-1312312 on Build GRCh38
rs371579977 Chr.1: 1312250-1312250 on Build GRCh38
rs974587340 Chr.1: 1312212-1312212 on Build GRCh38
rs1016019182 Chr.1: 1312207-1312207 on Build GRCh38
rs989738701 Chr.1: 1312262-1312262 on Build GRCh38
rs948085205 Chr.1: 1312122-1312122 on Build GRCh38
rs771029339 Chr.1: 1312126-1312126 on Build GRCh38
rs763231138 Chr.1: 1312204-1312205 on Build GRCh38
rs778572020 Chr.1: 1312274-1312274 on Build GRCh38
rs772758124 Chr.1: 1312284-1312284 on Build GRCh38
rs774539554 Chr.1: 1312205-1312206 on Build GRCh38
rs373564983 Chr.1: 1312216-1312216 on Build GRCh38
rs765446821 Chr.1: 1312248-1312248 on Build GRCh38
rs771555503 Chr.1: 1312282-1312282 on Build GRCh38
rs749071463 Chr.1: 1312267-1312267 on Build GRCh38
rs750273496 Chr.1: 1312197-1312198 on Build GRCh38
rs752605219 Chr.1: 1312198-1312199 on Build GRCh38
rs750300935 Chr.1: 1312272-1312273 on Build GRCh38
rs370137419 Chr.1: 1312296-1312296 on Build GRCh38
rs747481892 Chr.1: 1312290-1312290 on Build GRCh38
rs112135993 Chr.1: 1312152-1312152 on Build GRCh38
rs747584613 Chr.1: 1312200-1312201 on Build GRCh38
rs144812141 Chr.1: 1312199-1312200 on Build GRCh38
rs768395553 Chr.1: 1312215-1312215 on Build GRCh38
rs199818878 Chr.1: 1312261-1312261 on Build GRCh38
rs773703895 Chr.1: 1312234-1312234 on Build GRCh38
rs761226144 Chr.1: 1312239-1312239 on Build GRCh38
rs755276426 Chr.1: 1312195-1312195 on Build GRCh38
rs762277285 Chr.1: 1312161-1312161 on Build GRCh38
rs113643330 Chr.1: 1312225-1312225 on Build GRCh38
rs70949570 Chr.1: 1312208-1312209 on Build GRCh38
rs556901913 Chr.1: 1312291-1312291 on Build GRCh38
rs368932535 Chr.1: 1312147-1312147 on Build GRCh38
rs113412865 Chr.1: 1312175-1312175 on Build GRCh38
rs775853479 Chr.1: 1312300-1312300 on Build GRCh38
rs1055551604 Chr.1: 1312157-1312157 on Build GRCh38
rs201097701 Chr.1: 1312162-1312162 on Build GRCh38
rs887117870 Chr.1: 1312323-1312323 on Build GRCh38
rs753992813 Chr.1: 1312190-1312190 on Build GRCh38
rs201688337 Chr.1: 1312246-1312246 on Build GRCh38
rs746289153 Chr.1: 1312202-1312203 on Build GRCh38
rs767334657 Chr.1: 1312196-1312197 on Build GRCh38
rs868728948 Chr.1: 1312198-1312198 on Build GRCh38
rs761770529 Chr.1: 1312214-1312215 on Build GRCh38
rs755005813 Chr.1: 1312115-1312115 on Build GRCh38
rs747610411 Chr.1: 1312220-1312220 on Build GRCh38
rs754751339 Chr.1: 1312272-1312272 on Build GRCh38
rs916569478 Chr.1: 1312305-1312305 on Build GRCh38

View Full Product Details