Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29480118-29480383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN2
Entrez Gene ID
1124
Gene Name
chimerin 2
Gene Aliases
ARHGAP3, BCH, CHN2-3, RHOGAP3
UniGene
Hs.654611
Gene Chromosome Location
Chr.7: 29146547-29514335 on Build GRCh38
Transcripts
NM_001039936.2 NM_001293069.1 NM_001293070.1 NM_001293071.1 NM_001293072.1 NM_001293073.1 NM_001293075.1 NM_001293076.1 NM_001293077.1 NM_001293078.1 NM_001293079.1 NM_001293080.1 NM_004067.3 NR_120524.1 NR_120525.1 XM_011515105.2 XM_011515106.2 XM_011515107.2 XM_017011721.1 XM_017011722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3637810 c.879G>A p.K293K Chr.7: 29480356-29480356 on Build GRCh38
6497760 c.8C>A p.S3Y Chr.7: 29480118-29480118 on Build GRCh38
4979826 c.176C>G p.S59C Chr.7: 29480286-29480286 on Build GRCh38
5656550 c.847C>G p.H283D Chr.7: 29480324-29480324 on Build GRCh38
4964203 c.605T>G p.L202R Chr.7: 29480307-29480307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777546165 Chr.7: 29480224-29480224 on Build GRCh38
rs567620461 Chr.7: 29480217-29480217 on Build GRCh38
rs745379471 Chr.7: 29480326-29480326 on Build GRCh38
rs745463736 Chr.7: 29480240-29480240 on Build GRCh38
rs202070994 Chr.7: 29480237-29480237 on Build GRCh38
rs769478023 Chr.7: 29480338-29480338 on Build GRCh38
rs754827172 Chr.7: 29480213-29480213 on Build GRCh38
rs531399008 Chr.7: 29480262-29480262 on Build GRCh38
rs762443994 Chr.7: 29480186-29480186 on Build GRCh38
rs534685006 Chr.7: 29480218-29480218 on Build GRCh38
rs773777275 Chr.7: 29480356-29480356 on Build GRCh38
rs774848070 Chr.7: 29480251-29480251 on Build GRCh38
rs367953107 Chr.7: 29480212-29480212 on Build GRCh38
rs766731395 Chr.7: 29480209-29480209 on Build GRCh38
rs752352099 Chr.7: 29480192-29480192 on Build GRCh38
rs988712495 Chr.7: 29480268-29480268 on Build GRCh38
rs769425064 Chr.7: 29480245-29480245 on Build GRCh38
rs750560376 Chr.7: 29480203-29480203 on Build GRCh38
rs767909239 Chr.7: 29480187-29480187 on Build GRCh38
rs940209868 Chr.7: 29480324-29480324 on Build GRCh38
rs748568477 Chr.7: 29480352-29480352 on Build GRCh38
rs778441827 Chr.7: 29480281-29480283 on Build GRCh38
rs765000044 Chr.7: 29480146-29480146 on Build GRCh38
rs762645556 Chr.7: 29480293-29480293 on Build GRCh38
rs756772424 Chr.7: 29480322-29480322 on Build GRCh38
rs199586402 Chr.7: 29480271-29480271 on Build GRCh38
rs774849898 Chr.7: 29480163-29480163 on Build GRCh38
rs756130664 Chr.7: 29480204-29480204 on Build GRCh38
rs772289468 Chr.7: 29480353-29480353 on Build GRCh38
rs773547678 Chr.7: 29480279-29480279 on Build GRCh38
rs779551481 Chr.7: 29480348-29480348 on Build GRCh38
rs752051413 Chr.7: 29480322-29480324 on Build GRCh38
rs369197548 Chr.7: 29480318-29480318 on Build GRCh38
rs780736484 Chr.7: 29480323-29480323 on Build GRCh38
rs764372418 Chr.7: 29480162-29480162 on Build GRCh38
rs957366214 Chr.7: 29480232-29480232 on Build GRCh38
rs879172564 Chr.7: 29480280-29480280 on Build GRCh38
rs765522042 Chr.7: 29480298-29480298 on Build GRCh38
rs766502752 Chr.7: 29480281-29480281 on Build GRCh38
rs758075685 Chr.7: 29480317-29480317 on Build GRCh38
rs899190946 Chr.7: 29480179-29480179 on Build GRCh38
rs770388092 Chr.7: 29480236-29480236 on Build GRCh38
rs758397686 Chr.7: 29480222-29480222 on Build GRCh38
rs763336210 Chr.7: 29480150-29480150 on Build GRCh38
rs959933512 Chr.7: 29480266-29480266 on Build GRCh38
rs978841126 Chr.7: 29480295-29480296 on Build GRCh38
rs916128014 Chr.7: 29480238-29480238 on Build GRCh38
rs746551219 Chr.7: 29480227-29480227 on Build GRCh38
rs755316622 Chr.7: 29480294-29480294 on Build GRCh38
rs3750103 Chr.7: 29480313-29480313 on Build GRCh38
rs768119351 Chr.7: 29480276-29480276 on Build GRCh38
rs979935683 Chr.7: 29480231-29480231 on Build GRCh38
rs1033540100 Chr.7: 29480202-29480202 on Build GRCh38
rs761209183 Chr.7: 29480362-29480362 on Build GRCh38

View Full Product Details