Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144080313-144080586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC4
Entrez Gene ID
54512
Gene Name
exosome component 4
Gene Aliases
RRP41, RRP41A, Rrp41p, SKI6, Ski6p, hRrp41p, p12A
UniGene
Hs.632041
Gene Chromosome Location
Chr.8: 144064092-144080648 on Build GRCh38
Transcripts
NM_019037.2 XM_011517134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6657309 c.668_669ins14 p.D229fs*5 Chr.8: 144080531-144080532 on Build GRCh38
5510332 c.499G>A p.G167S Chr.8: 144080362-144080362 on Build GRCh38
1578579 c.610C>T p.R204W Chr.8: 144080473-144080473 on Build GRCh38
6657308 c.492C>T p.F164F Chr.8: 144080355-144080355 on Build GRCh38
6842759 c.524A>G p.H175R Chr.8: 144080387-144080387 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1132181 Chr.8: 144080494-144080494 on Build GRCh38
rs782033267 Chr.8: 144080420-144080420 on Build GRCh38
rs200725192 Chr.8: 144080482-144080482 on Build GRCh38
rs782353429 Chr.8: 144080542-144080542 on Build GRCh38
rs145830532 Chr.8: 144080339-144080339 on Build GRCh38
rs528426689 Chr.8: 144080469-144080469 on Build GRCh38
rs782147811 Chr.8: 144080428-144080428 on Build GRCh38
rs141844922 Chr.8: 144080355-144080355 on Build GRCh38
rs1132182 Chr.8: 144080497-144080497 on Build GRCh38
rs782559026 Chr.8: 144080366-144080366 on Build GRCh38
rs370639052 Chr.8: 144080472-144080472 on Build GRCh38
rs1003494401 Chr.8: 144080410-144080410 on Build GRCh38
rs369053542 Chr.8: 144080373-144080373 on Build GRCh38
rs201609181 Chr.8: 144080454-144080454 on Build GRCh38
rs148476615 Chr.8: 144080361-144080361 on Build GRCh38
rs141963829 Chr.8: 144080386-144080386 on Build GRCh38
rs781895679 Chr.8: 144080415-144080416 on Build GRCh38
rs556902394 Chr.8: 144080340-144080340 on Build GRCh38
rs367990298 Chr.8: 144080567-144080567 on Build GRCh38
rs1132175 Chr.8: 144080488-144080488 on Build GRCh38
rs782648570 Chr.8: 144080526-144080526 on Build GRCh38
rs782295020 Chr.8: 144080506-144080506 on Build GRCh38
rs376508817 Chr.8: 144080453-144080453 on Build GRCh38
rs781977410 Chr.8: 144080450-144080450 on Build GRCh38
rs145828514 Chr.8: 144080481-144080481 on Build GRCh38
rs141229810 Chr.8: 144080551-144080551 on Build GRCh38
rs200442280 Chr.8: 144080434-144080434 on Build GRCh38
rs782462065 Chr.8: 144080430-144080430 on Build GRCh38
rs782316494 Chr.8: 144080561-144080561 on Build GRCh38
rs782670087 Chr.8: 144080371-144080371 on Build GRCh38
rs373942429 Chr.8: 144080474-144080474 on Build GRCh38
rs782036452 Chr.8: 144080564-144080564 on Build GRCh38
rs377386138 Chr.8: 144080498-144080498 on Build GRCh38
rs1021824104 Chr.8: 144080527-144080527 on Build GRCh38
rs147555477 Chr.8: 144080376-144080376 on Build GRCh38
rs539358141 Chr.8: 144080377-144080377 on Build GRCh38
rs142724505 Chr.8: 144080528-144080528 on Build GRCh38
rs781799481 Chr.8: 144080362-144080362 on Build GRCh38
rs372756031 Chr.8: 144080350-144080350 on Build GRCh38
rs782817123 Chr.8: 144080460-144080460 on Build GRCh38
rs199867514 Chr.8: 144080510-144080510 on Build GRCh38
rs782220917 Chr.8: 144080560-144080560 on Build GRCh38

View Full Product Details