Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210203272-210203503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADL
Entrez Gene ID
33
Gene Name
acyl-CoA dehydrogenase, long chain
Gene Aliases
ACAD4, LCAD
UniGene
Hs.471277
Gene Chromosome Location
Chr.2: 210187990-210225491 on Build GRCh38
Transcripts
NM_001608.3 XM_005246517.4 XM_017003955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5346511 c.929C>T p.T310I Chr.2: 210203386-210203386 on Build GRCh38
6480180 c.889G>A p.D297N Chr.2: 210203426-210203426 on Build GRCh38
5346620 c.930C>G p.T310T Chr.2: 210203385-210203385 on Build GRCh38
6396803 c.874A>G p.R292G Chr.2: 210203441-210203441 on Build GRCh38
5346276 c.932G>T p.R311M Chr.2: 210203383-210203383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754366098 Chr.2: 210203453-210203453 on Build GRCh38
rs754063899 Chr.2: 210203444-210203444 on Build GRCh38
rs750962101 Chr.2: 210203405-210203405 on Build GRCh38
rs768043437 Chr.2: 210203375-210203375 on Build GRCh38
rs532924995 Chr.2: 210203383-210203384 on Build GRCh38
rs769217384 Chr.2: 210203329-210203329 on Build GRCh38
rs561311003 Chr.2: 210203382-210203383 on Build GRCh38
rs777751753 Chr.2: 210203306-210203306 on Build GRCh38
rs150159421 Chr.2: 210203411-210203411 on Build GRCh38
rs1016655243 Chr.2: 210203300-210203300 on Build GRCh38
rs745308856 Chr.2: 210203324-210203324 on Build GRCh38
rs764485489 Chr.2: 210203378-210203378 on Build GRCh38
rs761333691 Chr.2: 210203382-210203393 on Build GRCh38
rs771051970 Chr.2: 210203322-210203322 on Build GRCh38
rs143793918 Chr.2: 210203417-210203417 on Build GRCh38
rs549315531 Chr.2: 210203386-210203387 on Build GRCh38
rs779456934 Chr.2: 210203379-210203380 on Build GRCh38
rs767950958 Chr.2: 210203395-210203395 on Build GRCh38
rs749498544 Chr.2: 210203301-210203301 on Build GRCh38
rs755222295 Chr.2: 210203447-210203447 on Build GRCh38
rs377085604 Chr.2: 210203383-210203383 on Build GRCh38
rs538138862 Chr.2: 210203316-210203316 on Build GRCh38
rs759975624 Chr.2: 210203347-210203348 on Build GRCh38
rs866467886 Chr.2: 210203333-210203333 on Build GRCh38
rs775159045 Chr.2: 210203335-210203335 on Build GRCh38
rs546377607 Chr.2: 210203381-210203382 on Build GRCh38
rs776724966 Chr.2: 210203323-210203323 on Build GRCh38
rs1801204 Chr.2: 210203407-210203407 on Build GRCh38
rs762291639 Chr.2: 210203385-210203385 on Build GRCh38
rs1042213704 Chr.2: 210203471-210203471 on Build GRCh38
rs758194520 Chr.2: 210203303-210203303 on Build GRCh38
rs148069105 Chr.2: 210203376-210203376 on Build GRCh38
rs766421510 Chr.2: 210203421-210203421 on Build GRCh38

View Full Product Details