Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241505908-241506159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FH
Entrez Gene ID
2271
Gene Name
fumarate hydratase
Gene Aliases
FMRD, HLRCC, LRCC, MCL, MCUL1
UniGene
Hs.592490
Gene Chromosome Location
Chr.1: 241497557-241519785 on Build GRCh38
Transcripts
NM_000143.3 XM_011544132.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5943209 c.898_904+4delCTTACAGGTTA p.? Chr.1: 241505999-241506009 on Build GRCh38
679715 c.885A>G p.A295A Chr.1: 241506022-241506022 on Build GRCh38
5949782 c.890T>C p.V297A Chr.1: 241506017-241506017 on Build GRCh38
3864962 c.904+2T>G p.? Chr.1: 241506001-241506001 on Build GRCh38
308032 c.884C>T p.A295V Chr.1: 241506023-241506023 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145209119 Chr.1: 241505956-241505956 on Build GRCh38
rs567291615 Chr.1: 241506097-241506097 on Build GRCh38
rs781047980 Chr.1: 241505991-241505991 on Build GRCh38
rs113349873 Chr.1: 241506076-241506076 on Build GRCh38
rs34827919 Chr.1: 241505959-241505959 on Build GRCh38
rs754201668 Chr.1: 241505970-241505970 on Build GRCh38
rs786203177 Chr.1: 241506122-241506122 on Build GRCh38
rs863223986 Chr.1: 241506011-241506013 on Build GRCh38
rs759596203 Chr.1: 241506055-241506055 on Build GRCh38
rs863223985 Chr.1: 241506071-241506091 on Build GRCh38
rs147437099 Chr.1: 241506026-241506026 on Build GRCh38
rs779717143 Chr.1: 241505976-241505976 on Build GRCh38
rs935002190 Chr.1: 241506062-241506062 on Build GRCh38
rs200791185 Chr.1: 241506054-241506054 on Build GRCh38
rs387906545 Chr.1: 241506114-241506114 on Build GRCh38
rs863223969 Chr.1: 241506068-241506068 on Build GRCh38
rs863223984 Chr.1: 241506112-241506113 on Build GRCh38
rs199635833 Chr.1: 241506022-241506022 on Build GRCh38
rs759618470 Chr.1: 241505952-241505952 on Build GRCh38
rs758146903 Chr.1: 241505971-241505971 on Build GRCh38
rs772190176 Chr.1: 241506090-241506090 on Build GRCh38
rs372099505 Chr.1: 241506013-241506013 on Build GRCh38
rs771087739 Chr.1: 241506033-241506033 on Build GRCh38
rs12065955 Chr.1: 241505949-241505949 on Build GRCh38
rs775368701 Chr.1: 241506091-241506091 on Build GRCh38
rs764648497 Chr.1: 241506110-241506110 on Build GRCh38
rs572806212 Chr.1: 241505990-241505990 on Build GRCh38
rs200412958 Chr.1: 241506003-241506003 on Build GRCh38
rs764562737 Chr.1: 241505963-241505963 on Build GRCh38
rs61736558 Chr.1: 241506125-241506125 on Build GRCh38
rs779019570 Chr.1: 241506093-241506093 on Build GRCh38
rs863223981 Chr.1: 241506109-241506110 on Build GRCh38
rs368234566 Chr.1: 241506115-241506115 on Build GRCh38
rs199829765 Chr.1: 241506123-241506123 on Build GRCh38
rs863223999 Chr.1: 241506002-241506002 on Build GRCh38
rs201395553 Chr.1: 241506015-241506015 on Build GRCh38
rs377015873 Chr.1: 241506102-241506102 on Build GRCh38
rs754285771 Chr.1: 241506111-241506111 on Build GRCh38
rs202060616 Chr.1: 241506098-241506098 on Build GRCh38
rs111751087 Chr.1: 241505954-241505954 on Build GRCh38
rs751072171 Chr.1: 241505982-241505982 on Build GRCh38
rs749029092 Chr.1: 241506030-241506030 on Build GRCh38
rs863223970 Chr.1: 241506029-241506029 on Build GRCh38
rs933479705 Chr.1: 241506020-241506020 on Build GRCh38
rs761190882 Chr.1: 241505957-241505957 on Build GRCh38
rs145843819 Chr.1: 241506024-241506024 on Build GRCh38
rs774598788 Chr.1: 241506046-241506046 on Build GRCh38
rs1047260812 Chr.1: 241505936-241505936 on Build GRCh38
rs765997824 Chr.1: 241506119-241506119 on Build GRCh38

View Full Product Details