Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45405430-45405667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_130445.3

SNP Detail

SNP ID Chromosome Location
rs951456850 Chr.21: 45405506-45405506 on Build GRCh38
rs771434230 Chr.21: 45405507-45405507 on Build GRCh38
rs936261834 Chr.21: 45405631-45405631 on Build GRCh38
rs986441476 Chr.21: 45405561-45405561 on Build GRCh38
rs962625136 Chr.21: 45405539-45405539 on Build GRCh38
rs945129216 Chr.21: 45405568-45405568 on Build GRCh38
rs879434103 Chr.21: 45405455-45405455 on Build GRCh38
rs768235188 Chr.21: 45405517-45405517 on Build GRCh38
rs528787430 Chr.21: 45405494-45405494 on Build GRCh38
rs1056476113 Chr.21: 45405645-45405645 on Build GRCh38
rs762727400 Chr.21: 45405515-45405515 on Build GRCh38
rs1014332547 Chr.21: 45405481-45405481 on Build GRCh38
rs561528069 Chr.21: 45405483-45405483 on Build GRCh38
rs920921193 Chr.21: 45405544-45405544 on Build GRCh38
rs926146832 Chr.21: 45405607-45405607 on Build GRCh38
rs955070142 Chr.21: 45405553-45405553 on Build GRCh38
rs772492972 Chr.21: 45405463-45405463 on Build GRCh38
rs748662606 Chr.21: 45405460-45405460 on Build GRCh38
rs546242066 Chr.21: 45405588-45405588 on Build GRCh38
rs956513841 Chr.21: 45405557-45405557 on Build GRCh38
rs913629811 Chr.21: 45405567-45405567 on Build GRCh38
rs1055793223 Chr.21: 45405462-45405462 on Build GRCh38
rs775215088 Chr.21: 45405508-45405508 on Build GRCh38

View Full Product Details