Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 235744623-235744874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGAP1
Entrez Gene ID
116987
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 1
Gene Aliases
AGAP-1, CENTG2, GGAP1, cnt-g2
UniGene
Hs.435039
Gene Chromosome Location
Chr.2: 235494089-236131800 on Build GRCh38
Transcripts
NM_001037131.2 NM_001244888.1 NM_014914.4 XM_005246059.4 XM_006712234.3 XM_006712235.3 XM_006712237.3 XM_006712239.3 XM_011510547.2 XM_011510548.2 XM_011510549.2 XM_017003282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182178 c.415G>A p.A139T Chr.2: 235744716-235744716 on Build GRCh38
7128944 c.528G>A p.V176V Chr.2: 235744829-235744829 on Build GRCh38
6157525 c.479G>T p.S160I Chr.2: 235744780-235744780 on Build GRCh38
5396537 c.523C>T p.L175L Chr.2: 235744824-235744824 on Build GRCh38
720135 c.456C>G p.F152L Chr.2: 235744757-235744757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs538844406 Chr.2: 235744783-235744783 on Build GRCh38
rs17855721 Chr.2: 235744744-235744744 on Build GRCh38
rs758794882 Chr.2: 235744723-235744723 on Build GRCh38
rs763866531 Chr.2: 235744660-235744660 on Build GRCh38
rs752963873 Chr.2: 235744718-235744718 on Build GRCh38
rs779782500 Chr.2: 235744716-235744716 on Build GRCh38
rs1048563913 Chr.2: 235744738-235744738 on Build GRCh38
rs776822366 Chr.2: 235744651-235744651 on Build GRCh38
rs761391074 Chr.2: 235744706-235744706 on Build GRCh38
rs779138865 Chr.2: 235744681-235744691 on Build GRCh38
rs753542683 Chr.2: 235744797-235744797 on Build GRCh38
rs756230653 Chr.2: 235744818-235744818 on Build GRCh38
rs746030307 Chr.2: 235744682-235744684 on Build GRCh38
rs115617155 Chr.2: 235744648-235744648 on Build GRCh38
rs575855431 Chr.2: 235744843-235744843 on Build GRCh38
rs773937875 Chr.2: 235744686-235744686 on Build GRCh38
rs770964719 Chr.2: 235744739-235744739 on Build GRCh38
rs368517565 Chr.2: 235744762-235744762 on Build GRCh38
rs757710967 Chr.2: 235744808-235744808 on Build GRCh38
rs376740199 Chr.2: 235744777-235744777 on Build GRCh38
rs751552809 Chr.2: 235744666-235744666 on Build GRCh38
rs757425867 Chr.2: 235744674-235744676 on Build GRCh38
rs758678583 Chr.2: 235744798-235744798 on Build GRCh38
rs766087479 Chr.2: 235744807-235744807 on Build GRCh38
rs373927513 Chr.2: 235744656-235744656 on Build GRCh38
rs149414732 Chr.2: 235744786-235744786 on Build GRCh38
rs781296171 Chr.2: 235744740-235744740 on Build GRCh38
rs750072349 Chr.2: 235744710-235744710 on Build GRCh38
rs199790517 Chr.2: 235744851-235744851 on Build GRCh38
rs749589952 Chr.2: 235744846-235744846 on Build GRCh38
rs747025468 Chr.2: 235744649-235744649 on Build GRCh38
rs762837318 Chr.2: 235744659-235744659 on Build GRCh38
rs765881581 Chr.2: 235744795-235744795 on Build GRCh38
rs144494543 Chr.2: 235744805-235744805 on Build GRCh38
rs370523881 Chr.2: 235744653-235744653 on Build GRCh38
rs200676786 Chr.2: 235744764-235744764 on Build GRCh38
rs761885484 Chr.2: 235744776-235744776 on Build GRCh38
rs141449009 Chr.2: 235744770-235744770 on Build GRCh38
rs376672699 Chr.2: 235744834-235744834 on Build GRCh38
rs138458059 Chr.2: 235744715-235744715 on Build GRCh38
rs372718570 Chr.2: 235744809-235744809 on Build GRCh38
rs768537581 Chr.2: 235744848-235744848 on Build GRCh38
rs778196676 Chr.2: 235744725-235744725 on Build GRCh38
rs553037277 Chr.2: 235744763-235744763 on Build GRCh38
rs769867020 Chr.2: 235744751-235744751 on Build GRCh38
rs913594838 Chr.2: 235744824-235744824 on Build GRCh38
rs745864019 Chr.2: 235744815-235744815 on Build GRCh38
rs775098149 Chr.2: 235744657-235744657 on Build GRCh38
rs747575442 Chr.2: 235744726-235744726 on Build GRCh38
rs368843172 Chr.2: 235744791-235744791 on Build GRCh38
rs745982151 Chr.2: 235744748-235744748 on Build GRCh38
rs755702930 Chr.2: 235744804-235744804 on Build GRCh38
rs961658221 Chr.2: 235744737-235744737 on Build GRCh38

View Full Product Details