Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17420129-17420396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MVB12A
Entrez Gene ID
93343
Gene Name
multivesicular body subunit 12A
Gene Aliases
CFBP, FAM125A
UniGene
Hs.515243
Gene Chromosome Location
Chr.19: 17405686-17425339 on Build GRCh38
Transcripts
NM_001304547.1 NM_138401.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140370 c.100A>C p.T34P Chr.19: 17420322-17420322 on Build GRCh38
711250 c.159C>T p.F53F Chr.19: 17420381-17420381 on Build GRCh38
992793 c.113C>A p.A38E Chr.19: 17420335-17420335 on Build GRCh38
1206074 c.173C>A p.S58Y Chr.19: 17420395-17420395 on Build GRCh38
397007 c.138C>T p.F46F Chr.19: 17420360-17420360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778649038 Chr.19: 17420352-17420352 on Build GRCh38
rs202023401 Chr.19: 17420339-17420339 on Build GRCh38
rs755892412 Chr.19: 17420271-17420271 on Build GRCh38
rs147670744 Chr.19: 17420362-17420362 on Build GRCh38
rs376262904 Chr.19: 17420272-17420272 on Build GRCh38
rs755912420 Chr.19: 17420206-17420206 on Build GRCh38
rs780334293 Chr.19: 17420278-17420279 on Build GRCh38
rs746626401 Chr.19: 17420374-17420374 on Build GRCh38
rs369154468 Chr.19: 17420296-17420296 on Build GRCh38
rs751797077 Chr.19: 17420256-17420256 on Build GRCh38
rs762965710 Chr.19: 17420324-17420324 on Build GRCh38
rs754315810 Chr.19: 17420159-17420159 on Build GRCh38
rs775984858 Chr.19: 17420311-17420311 on Build GRCh38
rs749244237 Chr.19: 17420220-17420220 on Build GRCh38
rs771212419 Chr.19: 17420227-17420227 on Build GRCh38
rs766175288 Chr.19: 17420340-17420340 on Build GRCh38
rs1016046205 Chr.19: 17420150-17420150 on Build GRCh38
rs1054131261 Chr.19: 17420307-17420307 on Build GRCh38
rs764239160 Chr.19: 17420243-17420243 on Build GRCh38
rs745626383 Chr.19: 17420204-17420204 on Build GRCh38
rs778933470 Chr.19: 17420287-17420287 on Build GRCh38
rs753779719 Chr.19: 17420274-17420274 on Build GRCh38
rs1009212414 Chr.19: 17420160-17420160 on Build GRCh38
rs963102104 Chr.19: 17420154-17420154 on Build GRCh38
rs747621670 Chr.19: 17420293-17420293 on Build GRCh38
rs760063336 Chr.19: 17420303-17420303 on Build GRCh38
rs916216238 Chr.19: 17420161-17420161 on Build GRCh38
rs747780762 Chr.19: 17420223-17420223 on Build GRCh38
rs764219133 Chr.19: 17420334-17420334 on Build GRCh38
rs752695212 Chr.19: 17420356-17420356 on Build GRCh38
rs984382999 Chr.19: 17420211-17420211 on Build GRCh38
rs560187664 Chr.19: 17420309-17420309 on Build GRCh38
rs982083357 Chr.19: 17420166-17420166 on Build GRCh38
rs577744679 Chr.19: 17420193-17420193 on Build GRCh38
rs970794715 Chr.19: 17420162-17420162 on Build GRCh38
rs758656588 Chr.19: 17420201-17420201 on Build GRCh38
rs777180057 Chr.19: 17420233-17420233 on Build GRCh38
rs1003291164 Chr.19: 17420202-17420202 on Build GRCh38
rs935187186 Chr.19: 17420205-17420205 on Build GRCh38
rs752765335 Chr.19: 17420234-17420234 on Build GRCh38
rs768209524 Chr.19: 17420221-17420221 on Build GRCh38
rs773029690 Chr.19: 17420224-17420224 on Build GRCh38
rs777490142 Chr.19: 17420364-17420364 on Build GRCh38
rs776897000 Chr.19: 17420297-17420297 on Build GRCh38
rs751778674 Chr.19: 17420335-17420335 on Build GRCh38
rs765303621 Chr.19: 17420192-17420192 on Build GRCh38
rs985471650 Chr.19: 17420269-17420269 on Build GRCh38
rs1053598676 Chr.19: 17420157-17420157 on Build GRCh38
rs112659948 Chr.19: 17420171-17420171 on Build GRCh38
rs1040111652 Chr.19: 17420327-17420327 on Build GRCh38
rs371492135 Chr.19: 17420344-17420344 on Build GRCh38
rs763038997 Chr.19: 17420235-17420235 on Build GRCh38
rs779986939 Chr.19: 17420212-17420212 on Build GRCh38
rs540563524 Chr.19: 17420244-17420244 on Build GRCh38
rs755020873 Chr.19: 17420177-17420177 on Build GRCh38
rs1007821210 Chr.19: 17420250-17420250 on Build GRCh38

View Full Product Details