Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153930194-153930449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4B
Entrez Gene ID
9909
Gene Name
DENN domain containing 4B
Gene Aliases
KIAA0476
UniGene
Hs.744878
Gene Chromosome Location
Chr.1: 153929501-153946681 on Build GRCh38
Transcripts
NM_014856.2 XM_005245678.4 XM_005245679.2 XM_011510219.1 XM_011510220.2 XM_011510221.2 XM_017002993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4022677 c.4384C>A p.L1462M Chr.1: 153930404-153930404 on Build GRCh38
1134400 c.4421G>A p.R1474Q Chr.1: 153930367-153930367 on Build GRCh38
4022678 c.4048C>A p.L1350M Chr.1: 153930404-153930404 on Build GRCh38
6642433 c.4018G>A p.D1340N Chr.1: 153930434-153930434 on Build GRCh38
5434805 c.4399G>T p.G1467C Chr.1: 153930389-153930389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372005020 Chr.1: 153930390-153930390 on Build GRCh38
rs573948345 Chr.1: 153930367-153930367 on Build GRCh38
rs751531478 Chr.1: 153930365-153930365 on Build GRCh38
rs779916030 Chr.1: 153930364-153930364 on Build GRCh38
rs555769689 Chr.1: 153930263-153930263 on Build GRCh38
rs1012309117 Chr.1: 153930324-153930324 on Build GRCh38
rs758354105 Chr.1: 153930363-153930363 on Build GRCh38
rs772615819 Chr.1: 153930269-153930269 on Build GRCh38
rs774662631 Chr.1: 153930414-153930414 on Build GRCh38
rs771379877 Chr.1: 153930297-153930297 on Build GRCh38
rs752850099 Chr.1: 153930347-153930347 on Build GRCh38
rs748814708 Chr.1: 153930408-153930408 on Build GRCh38
rs1026698685 Chr.1: 153930223-153930223 on Build GRCh38
rs769616960 Chr.1: 153930387-153930387 on Build GRCh38
rs767881181 Chr.1: 153930337-153930337 on Build GRCh38
rs774770748 Chr.1: 153930313-153930313 on Build GRCh38
rs770568809 Chr.1: 153930410-153930410 on Build GRCh38
rs759874404 Chr.1: 153930328-153930328 on Build GRCh38
rs1053578102 Chr.1: 153930241-153930241 on Build GRCh38
rs982441457 Chr.1: 153930303-153930303 on Build GRCh38
rs922096092 Chr.1: 153930368-153930368 on Build GRCh38
rs763492051 Chr.1: 153930295-153930295 on Build GRCh38
rs747750051 Chr.1: 153930384-153930384 on Build GRCh38
rs995254705 Chr.1: 153930218-153930218 on Build GRCh38
rs764137281 Chr.1: 153930361-153930361 on Build GRCh38
rs201674736 Chr.1: 153930369-153930369 on Build GRCh38
rs760909337 Chr.1: 153930360-153930360 on Build GRCh38
rs893651550 Chr.1: 153930261-153930261 on Build GRCh38

View Full Product Details