Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 112778899-112779170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
-
UniGene
Hs.528051
Gene Chromosome Location
Chr.X: 112774877-112840946 on Build GRCh38
Transcripts
NM_001113490.1 NM_133265.2 XM_005262087.1 XM_005262090.1 XM_011530875.2 XM_017029289.1

SNP Detail

SNP ID Chromosome Location
rs759213175 Chr.X: 112779099-112779099 on Build GRCh38
rs923749335 Chr.X: 112778999-112778999 on Build GRCh38
rs188299748 Chr.X: 112778983-112778983 on Build GRCh38
rs769972836 Chr.X: 112779038-112779038 on Build GRCh38
rs780675984 Chr.X: 112779090-112779091 on Build GRCh38
rs773385598 Chr.X: 112779090-112779090 on Build GRCh38
rs755757620 Chr.X: 112779105-112779106 on Build GRCh38
rs747924574 Chr.X: 112779042-112779042 on Build GRCh38
rs373265985 Chr.X: 112779073-112779074 on Build GRCh38
rs757323299 Chr.X: 112779015-112779015 on Build GRCh38
rs773009609 Chr.X: 112779025-112779025 on Build GRCh38
rs781294453 Chr.X: 112779023-112779023 on Build GRCh38
rs765725477 Chr.X: 112779100-112779111 on Build GRCh38
rs764972866 Chr.X: 112779116-112779116 on Build GRCh38
rs778746629 Chr.X: 112779039-112779039 on Build GRCh38
rs146157772 Chr.X: 112779055-112779055 on Build GRCh38
rs778288036 Chr.X: 112778957-112778957 on Build GRCh38
rs897456274 Chr.X: 112779074-112779074 on Build GRCh38
rs12395302 Chr.X: 112778977-112778977 on Build GRCh38
rs759298835 Chr.X: 112779111-112779111 on Build GRCh38
rs138992506 Chr.X: 112779059-112779059 on Build GRCh38
rs149459069 Chr.X: 112779061-112779061 on Build GRCh38
rs147791784 Chr.X: 112779069-112779070 on Build GRCh38
rs753128850 Chr.X: 112779118-112779118 on Build GRCh38
rs764543196 Chr.X: 112779131-112779131 on Build GRCh38
rs771915548 Chr.X: 112779054-112779054 on Build GRCh38
rs941775503 Chr.X: 112779135-112779135 on Build GRCh38
rs763314264 Chr.X: 112779119-112779119 on Build GRCh38
rs968803867 Chr.X: 112778923-112778923 on Build GRCh38
rs761563277 Chr.X: 112778961-112778961 on Build GRCh38

View Full Product Details