Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 25679691-25679922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10A
Entrez Gene ID
57194
Gene Name
ATPase phospholipid transporting 10A (putative)
Gene Aliases
ATP10C, ATPVA, ATPVC
UniGene
Hs.638517
Gene Chromosome Location
Chr.15: 25672241-25865144 on Build GRCh38
Transcripts
NM_024490.3 XM_005268261.4 XM_011521826.2 XM_011521828.2 XM_011521829.2 XM_011521830.2 XM_017022436.1 XM_017022437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1245670 c.4062T>C p.S1354S Chr.15: 25679779-25679779 on Build GRCh38
4664129 c.3942C>A p.S1314S Chr.15: 25679899-25679899 on Build GRCh38
1163078 c.4133C>T p.P1378L Chr.15: 25679708-25679708 on Build GRCh38
7119773 c.3961C>A p.P1321T Chr.15: 25679880-25679880 on Build GRCh38
1245668 c.4029A>G p.T1343T Chr.15: 25679812-25679812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762484510 Chr.15: 25679828-25679828 on Build GRCh38
rs1053388386 Chr.15: 25679883-25679883 on Build GRCh38
rs142173664 Chr.15: 25679849-25679849 on Build GRCh38
rs371821622 Chr.15: 25679767-25679767 on Build GRCh38
rs752210077 Chr.15: 25679724-25679724 on Build GRCh38
rs766617998 Chr.15: 25679839-25679839 on Build GRCh38
rs780504341 Chr.15: 25679875-25679876 on Build GRCh38
rs769334065 Chr.15: 25679884-25679884 on Build GRCh38
rs1055136961 Chr.15: 25679852-25679852 on Build GRCh38
rs113319791 Chr.15: 25679762-25679762 on Build GRCh38
rs748862225 Chr.15: 25679900-25679900 on Build GRCh38
rs199978378 Chr.15: 25679761-25679761 on Build GRCh38
rs147816025 Chr.15: 25679855-25679855 on Build GRCh38
rs1051165903 Chr.15: 25679861-25679862 on Build GRCh38
rs770692793 Chr.15: 25679803-25679803 on Build GRCh38
rs112740057 Chr.15: 25679745-25679745 on Build GRCh38
rs892067754 Chr.15: 25679902-25679902 on Build GRCh38
rs201230742 Chr.15: 25679775-25679775 on Build GRCh38
rs751092237 Chr.15: 25679855-25679856 on Build GRCh38
rs746866255 Chr.15: 25679799-25679799 on Build GRCh38
rs1033392672 Chr.15: 25679788-25679788 on Build GRCh38
rs374849683 Chr.15: 25679713-25679713 on Build GRCh38
rs757180174 Chr.15: 25679864-25679864 on Build GRCh38
rs114736146 Chr.15: 25679833-25679833 on Build GRCh38
rs778885979 Chr.15: 25679768-25679768 on Build GRCh38
rs139061181 Chr.15: 25679740-25679740 on Build GRCh38
rs768204678 Chr.15: 25679829-25679829 on Build GRCh38
rs754188303 Chr.15: 25679840-25679840 on Build GRCh38
rs762735331 Chr.15: 25679847-25679850 on Build GRCh38
rs745618857 Chr.15: 25679872-25679872 on Build GRCh38
rs769437993 Chr.15: 25679816-25679816 on Build GRCh38
rs199994384 Chr.15: 25679718-25679718 on Build GRCh38
rs149100591 Chr.15: 25679814-25679814 on Build GRCh38
rs765147553 Chr.15: 25679835-25679841 on Build GRCh38
rs376143743 Chr.15: 25679848-25679848 on Build GRCh38
rs755173160 Chr.15: 25679844-25679844 on Build GRCh38
rs766126361 Chr.15: 25679853-25679854 on Build GRCh38
rs750223651 Chr.15: 25679840-25679840 on Build GRCh38
rs768077816 Chr.15: 25679739-25679739 on Build GRCh38
rs754533654 Chr.15: 25679858-25679859 on Build GRCh38
rs533802255 Chr.15: 25679853-25679853 on Build GRCh38
rs776387186 Chr.15: 25679813-25679813 on Build GRCh38
rs773839498 Chr.15: 25679832-25679832 on Build GRCh38
rs867481623 Chr.15: 25679793-25679793 on Build GRCh38
rs141308074 Chr.15: 25679865-25679865 on Build GRCh38
rs770637838 Chr.15: 25679714-25679714 on Build GRCh38
rs200297626 Chr.15: 25679820-25679820 on Build GRCh38
rs777738173 Chr.15: 25679856-25679856 on Build GRCh38
rs762431612 Chr.15: 25679738-25679738 on Build GRCh38
rs369977237 Chr.15: 25679858-25679858 on Build GRCh38
rs116163236 Chr.15: 25679786-25679786 on Build GRCh38
rs775275172 Chr.15: 25679888-25679888 on Build GRCh38
rs748172500 Chr.15: 25679774-25679774 on Build GRCh38
rs777686800 Chr.15: 25679789-25679789 on Build GRCh38

View Full Product Details