Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184182768-184183041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2M1
Entrez Gene ID
1173
Gene Name
adaptor related protein complex 2 mu 1 subunit
Gene Aliases
AP50, CLAPM1, mu2
UniGene
Hs.518460
Gene Chromosome Location
Chr.3: 184174846-184184091 on Build GRCh38
Transcripts
NM_001025205.1 NM_001311198.1 NM_004068.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
729582 c.1102G>T p.A368S Chr.3: 184182797-184182797 on Build GRCh38
4115672 c.1079G>A p.G360D Chr.3: 184182774-184182774 on Build GRCh38
6601630 c.1143G>A p.W381* Chr.3: 184182838-184182838 on Build GRCh38
6479781 c.1134G>C p.K378N Chr.3: 184182829-184182829 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773364939 Chr.3: 184182823-184182823 on Build GRCh38
rs746961969 Chr.3: 184182957-184182957 on Build GRCh38
rs774676330 Chr.3: 184183005-184183005 on Build GRCh38
rs746214925 Chr.3: 184183006-184183006 on Build GRCh38
rs760249807 Chr.3: 184182898-184182898 on Build GRCh38
rs749475650 Chr.3: 184182972-184182972 on Build GRCh38
rs1803438 Chr.3: 184182803-184182803 on Build GRCh38
rs371726224 Chr.3: 184182914-184182914 on Build GRCh38
rs939814381 Chr.3: 184182911-184182911 on Build GRCh38
rs541722787 Chr.3: 184182997-184182997 on Build GRCh38
rs902011278 Chr.3: 184182797-184182797 on Build GRCh38
rs776207361 Chr.3: 184182897-184182897 on Build GRCh38
rs758276552 Chr.3: 184182947-184182947 on Build GRCh38
rs843366 Chr.3: 184182892-184182892 on Build GRCh38
rs777860540 Chr.3: 184182964-184182964 on Build GRCh38
rs368179342 Chr.3: 184182909-184182909 on Build GRCh38
rs753467174 Chr.3: 184182934-184182934 on Build GRCh38
rs779949117 Chr.3: 184182950-184182950 on Build GRCh38
rs867598792 Chr.3: 184182933-184182933 on Build GRCh38
rs541840372 Chr.3: 184182890-184182890 on Build GRCh38
rs780004232 Chr.3: 184182796-184182796 on Build GRCh38
rs759996213 Chr.3: 184182880-184182880 on Build GRCh38
rs1035228686 Chr.3: 184182969-184182969 on Build GRCh38
rs1030332157 Chr.3: 184182986-184182986 on Build GRCh38
rs774599365 Chr.3: 184182879-184182879 on Build GRCh38
rs1014909343 Chr.3: 184182916-184182916 on Build GRCh38
rs368284020 Chr.3: 184182790-184182790 on Build GRCh38
rs750151113 Chr.3: 184182944-184182944 on Build GRCh38
rs1007823094 Chr.3: 184183004-184183004 on Build GRCh38
rs768647913 Chr.3: 184182977-184182979 on Build GRCh38
rs749476004 Chr.3: 184182847-184182847 on Build GRCh38
rs748219382 Chr.3: 184182804-184182804 on Build GRCh38
rs997962942 Chr.3: 184182961-184182961 on Build GRCh38
rs745780607 Chr.3: 184182787-184182787 on Build GRCh38
rs756119080 Chr.3: 184182959-184182959 on Build GRCh38
rs960579747 Chr.3: 184183013-184183013 on Build GRCh38
rs769916005 Chr.3: 184182818-184182818 on Build GRCh38
rs994118879 Chr.3: 184182955-184182955 on Build GRCh38
rs843365 Chr.3: 184182941-184182941 on Build GRCh38
rs764987371 Chr.3: 184182942-184182942 on Build GRCh38
rs771038660 Chr.3: 184182988-184182988 on Build GRCh38
rs201012736 Chr.3: 184182850-184182850 on Build GRCh38

View Full Product Details