Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48639594-48639861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA11
Entrez Gene ID
770
Gene Name
carbonic anhydrase 11
Gene Aliases
CARPX1
UniGene
Hs.428446
Gene Chromosome Location
Chr.19: 48637942-48646312 on Build GRCh38
Transcripts
NM_001217.4 NR_136241.1 NR_004401.2

Gene Symbol
SEC1P
Entrez Gene ID
653677
Gene Name
secretory blood group 1, pseudogene
Gene Aliases
SEC1
UniGene
Hs.443160
Gene Chromosome Location
Chr.19: 48638039-48682245 on Build GRCh38
Transcripts
NM_001217.4 NR_136241.1 NR_004401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3107738 c.526C>T p.R176C Chr.19: 48639829-48639829 on Build GRCh38
439918 c.558C>G p.L186L Chr.19: 48639797-48639797 on Build GRCh38
3672963 c.526C>A p.R176S Chr.19: 48639829-48639829 on Build GRCh38
3729730 c.541C>A p.L181M Chr.19: 48639814-48639814 on Build GRCh38
3107737 c.527G>A p.R176H Chr.19: 48639828-48639828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754468877 Chr.19: 48639809-48639809 on Build GRCh38
rs776763526 Chr.19: 48639765-48639765 on Build GRCh38
rs754442009 Chr.19: 48639628-48639628 on Build GRCh38
rs950494853 Chr.19: 48639618-48639618 on Build GRCh38
rs755953372 Chr.19: 48639645-48639645 on Build GRCh38
rs556886440 Chr.19: 48639676-48639676 on Build GRCh38
rs969173439 Chr.19: 48639632-48639632 on Build GRCh38
rs755761012 Chr.19: 48639838-48639838 on Build GRCh38
rs755006103 Chr.19: 48639782-48639782 on Build GRCh38
rs766042493 Chr.19: 48639617-48639617 on Build GRCh38
rs766996127 Chr.19: 48639788-48639788 on Build GRCh38
rs987856413 Chr.19: 48639824-48639824 on Build GRCh38
rs770843857 Chr.19: 48639665-48639665 on Build GRCh38
rs914151921 Chr.19: 48639700-48639700 on Build GRCh38
rs749291748 Chr.19: 48639653-48639653 on Build GRCh38
rs780739372 Chr.19: 48639642-48639642 on Build GRCh38
rs761486466 Chr.19: 48639697-48639697 on Build GRCh38
rs754982944 Chr.19: 48639620-48639620 on Build GRCh38
rs2303147 Chr.19: 48639768-48639768 on Build GRCh38
rs752332285 Chr.19: 48639827-48639827 on Build GRCh38
rs768745014 Chr.19: 48639753-48639753 on Build GRCh38
rs376281824 Chr.19: 48639648-48639648 on Build GRCh38
rs1016402001 Chr.19: 48639661-48639661 on Build GRCh38
rs774290568 Chr.19: 48639742-48639742 on Build GRCh38
rs781385438 Chr.19: 48639708-48639708 on Build GRCh38
rs946751825 Chr.19: 48639707-48639707 on Build GRCh38
rs754031869 Chr.19: 48639814-48639814 on Build GRCh38
rs928166771 Chr.19: 48639643-48639643 on Build GRCh38
rs1052658190 Chr.19: 48639675-48639675 on Build GRCh38
rs773501800 Chr.19: 48639781-48639781 on Build GRCh38
rs202241268 Chr.19: 48639819-48639819 on Build GRCh38
rs541610646 Chr.19: 48639829-48639829 on Build GRCh38
rs901036181 Chr.19: 48639815-48639815 on Build GRCh38
rs751441442 Chr.19: 48639679-48639681 on Build GRCh38
rs747787044 Chr.19: 48639644-48639644 on Build GRCh38
rs564861464 Chr.19: 48639726-48639726 on Build GRCh38
rs752065041 Chr.19: 48639799-48639799 on Build GRCh38
rs200972928 Chr.19: 48639745-48639745 on Build GRCh38
rs34019493 Chr.19: 48639667-48639668 on Build GRCh38
rs749755530 Chr.19: 48639780-48639780 on Build GRCh38
rs954758614 Chr.19: 48639785-48639785 on Build GRCh38
rs748594016 Chr.19: 48639724-48639724 on Build GRCh38

View Full Product Details