Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35465966-35466239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP250
Entrez Gene ID
11190
Gene Name
centrosomal protein 250
Gene Aliases
C-NAP1, CEP2, CNAP1
UniGene
Hs.443976
Gene Chromosome Location
Chr.20: 35455138-35517531 on Build GRCh38
Transcripts
NM_001318219.1 NM_007186.5 XM_005260262.4 XM_005260263.4 XM_005260264.4 XM_006723690.3 XM_006723691.1 XM_006723692.3 XM_006723693.3 XM_006723694.3 XM_011528517.2 XM_011528518.2 XM_017027617.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5755566 c.436A>G p.S146G Chr.20: 35466148-35466148 on Build GRCh38
5755567 c.437G>T p.S146I Chr.20: 35466149-35466149 on Build GRCh38
7166580 c.348G>C p.V116V Chr.20: 35466060-35466060 on Build GRCh38
3911183 c.439C>G p.R147G Chr.20: 35466151-35466151 on Build GRCh38
723910 c.327G>T p.R109S Chr.20: 35466039-35466039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988843590 Chr.20: 35466056-35466056 on Build GRCh38
rs773059939 Chr.20: 35466025-35466025 on Build GRCh38
rs545221768 Chr.20: 35466048-35466048 on Build GRCh38
rs763474027 Chr.20: 35466158-35466158 on Build GRCh38
rs779109342 Chr.20: 35466140-35466140 on Build GRCh38
rs770739949 Chr.20: 35466162-35466162 on Build GRCh38
rs769369386 Chr.20: 35466123-35466123 on Build GRCh38
rs764781875 Chr.20: 35466178-35466178 on Build GRCh38
rs772076350 Chr.20: 35466152-35466152 on Build GRCh38
rs757895388 Chr.20: 35466066-35466066 on Build GRCh38
rs759279138 Chr.20: 35466175-35466175 on Build GRCh38
rs377314862 Chr.20: 35466069-35466069 on Build GRCh38
rs6060428 Chr.20: 35466038-35466038 on Build GRCh38
rs756657396 Chr.20: 35466216-35466216 on Build GRCh38
rs776162097 Chr.20: 35466124-35466124 on Build GRCh38
rs368777120 Chr.20: 35466078-35466078 on Build GRCh38
rs781762969 Chr.20: 35466074-35466074 on Build GRCh38
rs990022541 Chr.20: 35465999-35465999 on Build GRCh38
rs753546802 Chr.20: 35466039-35466039 on Build GRCh38
rs775802794 Chr.20: 35466013-35466013 on Build GRCh38
rs147554271 Chr.20: 35466096-35466096 on Build GRCh38
rs375458172 Chr.20: 35466035-35466035 on Build GRCh38
rs955984907 Chr.20: 35465992-35465992 on Build GRCh38
rs375906452 Chr.20: 35466173-35466173 on Build GRCh38
rs752316298 Chr.20: 35466193-35466193 on Build GRCh38
rs941233169 Chr.20: 35466151-35466151 on Build GRCh38
rs749565417 Chr.20: 35466111-35466111 on Build GRCh38
rs763638652 Chr.20: 35466207-35466207 on Build GRCh38
rs200952347 Chr.20: 35466215-35466215 on Build GRCh38
rs780242737 Chr.20: 35466097-35466097 on Build GRCh38
rs760471750 Chr.20: 35466030-35466030 on Build GRCh38
rs761861984 Chr.20: 35466008-35466008 on Build GRCh38
rs373038487 Chr.20: 35466157-35466157 on Build GRCh38
rs898185757 Chr.20: 35466187-35466187 on Build GRCh38
rs373750157 Chr.20: 35466000-35466000 on Build GRCh38
rs372185386 Chr.20: 35466011-35466011 on Build GRCh38
rs768812496 Chr.20: 35466003-35466003 on Build GRCh38
rs140783729 Chr.20: 35466194-35466194 on Build GRCh38
rs1056612522 Chr.20: 35466179-35466179 on Build GRCh38
rs1032953546 Chr.20: 35466023-35466023 on Build GRCh38
rs549020994 Chr.20: 35466149-35466149 on Build GRCh38
rs367899516 Chr.20: 35466006-35466006 on Build GRCh38
rs911048979 Chr.20: 35466114-35466114 on Build GRCh38
rs764839938 Chr.20: 35466063-35466063 on Build GRCh38
rs369894925 Chr.20: 35466065-35466065 on Build GRCh38
rs750893887 Chr.20: 35466082-35466082 on Build GRCh38

View Full Product Details