Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 64695462-64695733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM159B
Entrez Gene ID
100132916
Gene Name
family with sequence similarity 159 member B
Gene Aliases
-
UniGene
Hs.591776
Gene Chromosome Location
Chr.5: 64690308-64718190 on Build GRCh38
Transcripts
NM_001164442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3828275 c.204G>C p.L68F Chr.5: 64695519-64695519 on Build GRCh38
4879876 c.229G>T p.V77F Chr.5: 64695544-64695544 on Build GRCh38
3941354 c.285G>A p.T95T Chr.5: 64695600-64695600 on Build GRCh38
4591316 c.331G>C p.A111P Chr.5: 64695646-64695646 on Build GRCh38
7428538 c.277C>T p.L93L Chr.5: 64695592-64695592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554141682 Chr.5: 64695502-64695502 on Build GRCh38
rs201615430 Chr.5: 64695599-64695599 on Build GRCh38
rs544886348 Chr.5: 64695689-64695689 on Build GRCh38
rs903297612 Chr.5: 64695512-64695512 on Build GRCh38
rs2305962 Chr.5: 64695646-64695646 on Build GRCh38
rs1025979682 Chr.5: 64695532-64695532 on Build GRCh38
rs16893062 Chr.5: 64695702-64695702 on Build GRCh38
rs1033350596 Chr.5: 64695647-64695647 on Build GRCh38
rs773058029 Chr.5: 64695641-64695641 on Build GRCh38
rs202064052 Chr.5: 64695625-64695625 on Build GRCh38
rs940486768 Chr.5: 64695692-64695692 on Build GRCh38
rs369208300 Chr.5: 64695605-64695605 on Build GRCh38
rs745867780 Chr.5: 64695528-64695528 on Build GRCh38
rs780659179 Chr.5: 64695591-64695591 on Build GRCh38
rs574012886 Chr.5: 64695544-64695544 on Build GRCh38
rs275830 Chr.5: 64695685-64695685 on Build GRCh38
rs563854983 Chr.5: 64695600-64695600 on Build GRCh38
rs1008675975 Chr.5: 64695569-64695569 on Build GRCh38
rs536795941 Chr.5: 64695645-64695645 on Build GRCh38
rs374375151 Chr.5: 64695553-64695553 on Build GRCh38
rs957319536 Chr.5: 64695657-64695657 on Build GRCh38
rs765914175 Chr.5: 64695660-64695660 on Build GRCh38

View Full Product Details