Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 237324675-237324948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A3
Entrez Gene ID
1293
Gene Name
collagen type VI alpha 3 chain
Gene Aliases
BTHLM1, DYT27, UCMD1
UniGene
Hs.233240
Gene Chromosome Location
Chr.2: 237324012-237434822 on Build GRCh38
Transcripts
NM_004369.3 NM_057166.4 NM_057167.3 XM_005246065.1 XM_005246066.1 XM_006712253.1 XM_011510574.1 XM_017003303.1 XM_017003304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4092628 c.9499G>A p.A3167T Chr.2: 237324809-237324809 on Build GRCh38
477132 c.9519T>A p.S3173R Chr.2: 237324789-237324789 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142546062 Chr.2: 237324816-237324816 on Build GRCh38
rs200934970 Chr.2: 237324843-237324843 on Build GRCh38
rs943987290 Chr.2: 237324915-237324915 on Build GRCh38
rs1035329885 Chr.2: 237324697-237324697 on Build GRCh38
rs1015710999 Chr.2: 237324712-237324712 on Build GRCh38
rs779061806 Chr.2: 237324910-237324910 on Build GRCh38
rs779220948 Chr.2: 237324827-237324827 on Build GRCh38
rs143856219 Chr.2: 237324836-237324836 on Build GRCh38
rs534981873 Chr.2: 237324857-237324857 on Build GRCh38
rs113696317 Chr.2: 237324873-237324873 on Build GRCh38
rs537511128 Chr.2: 237324809-237324809 on Build GRCh38
rs766439690 Chr.2: 237324823-237324823 on Build GRCh38
rs773708184 Chr.2: 237324861-237324861 on Build GRCh38
rs912591267 Chr.2: 237324794-237324794 on Build GRCh38
rs13032404 Chr.2: 237324840-237324840 on Build GRCh38
rs912532163 Chr.2: 237324924-237324924 on Build GRCh38
rs746487382 Chr.2: 237324772-237324772 on Build GRCh38
rs188002584 Chr.2: 237324723-237324723 on Build GRCh38
rs752558732 Chr.2: 237324727-237324727 on Build GRCh38
rs568632361 Chr.2: 237324800-237324800 on Build GRCh38
rs367899223 Chr.2: 237324820-237324820 on Build GRCh38
rs778508222 Chr.2: 237324754-237324754 on Build GRCh38
rs766395668 Chr.2: 237324735-237324737 on Build GRCh38
rs1008454322 Chr.2: 237324804-237324804 on Build GRCh38
rs769626058 Chr.2: 237324874-237324874 on Build GRCh38
rs375729697 Chr.2: 237324752-237324752 on Build GRCh38
rs370704105 Chr.2: 237324835-237324835 on Build GRCh38
rs752501337 Chr.2: 237324831-237324831 on Build GRCh38
rs756223167 Chr.2: 237324746-237324746 on Build GRCh38
rs748341458 Chr.2: 237324912-237324912 on Build GRCh38
rs4663722 Chr.2: 237324767-237324767 on Build GRCh38
rs150376179 Chr.2: 237324810-237324810 on Build GRCh38
rs755099760 Chr.2: 237324825-237324825 on Build GRCh38
rs886043555 Chr.2: 237324819-237324819 on Build GRCh38
rs1001587285 Chr.2: 237324696-237324696 on Build GRCh38
rs112123472 Chr.2: 237324830-237324830 on Build GRCh38
rs985554048 Chr.2: 237324917-237324917 on Build GRCh38
rs372371502 Chr.2: 237324858-237324858 on Build GRCh38
rs374024399 Chr.2: 237324821-237324821 on Build GRCh38
rs749535250 Chr.2: 237324751-237324751 on Build GRCh38
rs571172160 Chr.2: 237324771-237324771 on Build GRCh38
rs776342807 Chr.2: 237324801-237324801 on Build GRCh38
rs374558518 Chr.2: 237324722-237324722 on Build GRCh38
rs375892263 Chr.2: 237324864-237324865 on Build GRCh38
rs148183839 Chr.2: 237324784-237324784 on Build GRCh38
rs758310256 Chr.2: 237324834-237324834 on Build GRCh38
rs768032349 Chr.2: 237324730-237324730 on Build GRCh38
rs957142857 Chr.2: 237324826-237324826 on Build GRCh38
rs780508651 Chr.2: 237324842-237324842 on Build GRCh38
rs775160702 Chr.2: 237324850-237324850 on Build GRCh38
rs759043836 Chr.2: 237324803-237324803 on Build GRCh38
rs184617787 Chr.2: 237324824-237324824 on Build GRCh38
rs780296574 Chr.2: 237324747-237324747 on Build GRCh38
rs983186196 Chr.2: 237324711-237324711 on Build GRCh38
rs932697544 Chr.2: 237324897-237324897 on Build GRCh38
rs920003343 Chr.2: 237324876-237324876 on Build GRCh38
rs959767840 Chr.2: 237324698-237324698 on Build GRCh38
rs920132941 Chr.2: 237324753-237324753 on Build GRCh38
rs750460015 Chr.2: 237324731-237324731 on Build GRCh38

View Full Product Details