Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40140684-40140921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASC3
Entrez Gene ID
22794
Gene Name
cancer susceptibility candidate 3
Gene Aliases
BTZ, MLN51
UniGene
Hs.743287
Gene Chromosome Location
Chr.17: 40140254-40172178 on Build GRCh38
Transcripts
NM_007359.4 XM_005257163.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6374755 c.157G>T p.G53* Chr.17: 40140705-40140705 on Build GRCh38
5687544 c.228G>A p.E76E Chr.17: 40140776-40140776 on Build GRCh38
6376267 c.164G>A p.R55Q Chr.17: 40140712-40140712 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746942151 Chr.17: 40140739-40140739 on Build GRCh38
rs1051363672 Chr.17: 40140808-40140808 on Build GRCh38
rs1003235427 Chr.17: 40140805-40140805 on Build GRCh38
rs959123078 Chr.17: 40140812-40140812 on Build GRCh38
rs558372528 Chr.17: 40140875-40140875 on Build GRCh38
rs1025011193 Chr.17: 40140729-40140729 on Build GRCh38
rs778663672 Chr.17: 40140723-40140723 on Build GRCh38
rs531575982 Chr.17: 40140754-40140754 on Build GRCh38
rs927566160 Chr.17: 40140872-40140872 on Build GRCh38
rs779698182 Chr.17: 40140708-40140708 on Build GRCh38
rs771835371 Chr.17: 40140727-40140727 on Build GRCh38
rs777582439 Chr.17: 40140728-40140728 on Build GRCh38
rs945143379 Chr.17: 40140793-40140793 on Build GRCh38
rs778338353 Chr.17: 40140787-40140787 on Build GRCh38
rs543731181 Chr.17: 40140768-40140768 on Build GRCh38
rs532751417 Chr.17: 40140789-40140789 on Build GRCh38
rs768144459 Chr.17: 40140805-40140805 on Build GRCh38
rs367657738 Chr.17: 40140874-40140874 on Build GRCh38
rs1027042678 Chr.17: 40140721-40140721 on Build GRCh38
rs773759957 Chr.17: 40140809-40140834 on Build GRCh38
rs908044757 Chr.17: 40140718-40140718 on Build GRCh38
rs748293528 Chr.17: 40140829-40140829 on Build GRCh38
rs1038893730 Chr.17: 40140849-40140849 on Build GRCh38
rs767990289 Chr.17: 40140762-40140762 on Build GRCh38
rs898959999 Chr.17: 40140716-40140716 on Build GRCh38
rs906190516 Chr.17: 40140800-40140800 on Build GRCh38
rs913627525 Chr.17: 40140792-40140793 on Build GRCh38
rs1045996173 Chr.17: 40140869-40140869 on Build GRCh38
rs774654065 Chr.17: 40140755-40140755 on Build GRCh38
rs770909452 Chr.17: 40140742-40140742 on Build GRCh38
rs747851575 Chr.17: 40140726-40140726 on Build GRCh38
rs753403484 Chr.17: 40140712-40140712 on Build GRCh38
rs565408472 Chr.17: 40140788-40140788 on Build GRCh38
rs766998291 Chr.17: 40140769-40140769 on Build GRCh38
rs973143783 Chr.17: 40140816-40140816 on Build GRCh38
rs890153310 Chr.17: 40140827-40140827 on Build GRCh38
rs887188563 Chr.17: 40140887-40140887 on Build GRCh38
rs1033831586 Chr.17: 40140810-40140810 on Build GRCh38
rs547629691 Chr.17: 40140801-40140801 on Build GRCh38
rs763098180 Chr.17: 40140749-40140749 on Build GRCh38
rs746704281 Chr.17: 40140765-40140766 on Build GRCh38
rs754524245 Chr.17: 40140720-40140720 on Build GRCh38
rs566252028 Chr.17: 40140861-40140861 on Build GRCh38
rs899091919 Chr.17: 40140856-40140856 on Build GRCh38
rs1054086902 Chr.17: 40140886-40140886 on Build GRCh38
rs1027404174 Chr.17: 40140839-40140839 on Build GRCh38
rs943041997 Chr.17: 40140848-40140848 on Build GRCh38
rs750877718 Chr.17: 40140764-40140764 on Build GRCh38
rs775370619 Chr.17: 40140747-40140747 on Build GRCh38
rs761989441 Chr.17: 40140756-40140756 on Build GRCh38

View Full Product Details