Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 37737301-37737556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNBL1
Entrez Gene ID
56259
Gene Name
catenin beta like 1
Gene Aliases
C20orf33, NAP, P14L, PP8304, dJ633O20.1
UniGene
Hs.472667
Gene Chromosome Location
Chr.20: 37693955-37872129 on Build GRCh38
Transcripts
NM_001281495.1 NM_030877.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5669120 c.255G>A p.M85I Chr.20: 37737413-37737413 on Build GRCh38
347975 c.273G>C p.K91N Chr.20: 37737431-37737431 on Build GRCh38
6249754 c.244G>T p.V82L Chr.20: 37737402-37737402 on Build GRCh38
1411600 c.243A>G p.S81S Chr.20: 37737401-37737401 on Build GRCh38
4675402 c.289C>T p.Q97* Chr.20: 37737447-37737447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745310323 Chr.20: 37737456-37737456 on Build GRCh38
rs11549311 Chr.20: 37737437-37737437 on Build GRCh38
rs766443965 Chr.20: 37737350-37737350 on Build GRCh38
rs767893449 Chr.20: 37737528-37737528 on Build GRCh38
rs761829534 Chr.20: 37737508-37737508 on Build GRCh38
rs566341554 Chr.20: 37737515-37737515 on Build GRCh38
rs768607112 Chr.20: 37737501-37737501 on Build GRCh38
rs374758112 Chr.20: 37737357-37737357 on Build GRCh38
rs761621326 Chr.20: 37737329-37737329 on Build GRCh38
rs76902575 Chr.20: 37737524-37737524 on Build GRCh38
rs200788171 Chr.20: 37737525-37737525 on Build GRCh38
rs991704511 Chr.20: 37737478-37737478 on Build GRCh38
rs781506333 Chr.20: 37737420-37737420 on Build GRCh38
rs756531288 Chr.20: 37737427-37737427 on Build GRCh38
rs779914746 Chr.20: 37737458-37737458 on Build GRCh38
rs765404606 Chr.20: 37737337-37737337 on Build GRCh38
rs202008237 Chr.20: 37737451-37737451 on Build GRCh38
rs144576870 Chr.20: 37737457-37737457 on Build GRCh38
rs533034446 Chr.20: 37737366-37737366 on Build GRCh38
rs372060016 Chr.20: 37737504-37737504 on Build GRCh38
rs148717643 Chr.20: 37737439-37737439 on Build GRCh38
rs372114272 Chr.20: 37737340-37737340 on Build GRCh38
rs780568750 Chr.20: 37737404-37737405 on Build GRCh38
rs368200793 Chr.20: 37737487-37737487 on Build GRCh38
rs548482768 Chr.20: 37737351-37737351 on Build GRCh38
rs753021929 Chr.20: 37737529-37737529 on Build GRCh38
rs753254698 Chr.20: 37737422-37737422 on Build GRCh38
rs915762532 Chr.20: 37737500-37737500 on Build GRCh38
rs769577600 Chr.20: 37737511-37737511 on Build GRCh38
rs766432967 Chr.20: 37737517-37737517 on Build GRCh38

View Full Product Details