Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 185747254-185747521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
-
UniGene
Hs.98025
Gene Chromosome Location
Chr.2: 185738895-185833289 on Build GRCh38
Transcripts
NM_173651.3

SNP Detail

SNP ID Chromosome Location
rs368240698 Chr.2: 185747298-185747298 on Build GRCh38
rs939875634 Chr.2: 185747411-185747411 on Build GRCh38
rs777048411 Chr.2: 185747387-185747387 on Build GRCh38
rs747440726 Chr.2: 185747324-185747324 on Build GRCh38
rs926694612 Chr.2: 185747323-185747323 on Build GRCh38
rs547138962 Chr.2: 185747498-185747498 on Build GRCh38
rs561989893 Chr.2: 185747306-185747306 on Build GRCh38
rs933142132 Chr.2: 185747348-185747348 on Build GRCh38
rs528618028 Chr.2: 185747485-185747485 on Build GRCh38
rs1057189247 Chr.2: 185747459-185747459 on Build GRCh38
rs745401205 Chr.2: 185747326-185747326 on Build GRCh38
rs996494796 Chr.2: 185747484-185747484 on Build GRCh38
rs77069614 Chr.2: 185747434-185747434 on Build GRCh38
rs905850173 Chr.2: 185747428-185747428 on Build GRCh38
rs73980266 Chr.2: 185747365-185747365 on Build GRCh38
rs1043030112 Chr.2: 185747419-185747419 on Build GRCh38
rs980758814 Chr.2: 185747285-185747285 on Build GRCh38
rs913116238 Chr.2: 185747356-185747356 on Build GRCh38
rs1056295651 Chr.2: 185747349-185747349 on Build GRCh38
rs1052950107 Chr.2: 185747354-185747354 on Build GRCh38
rs531131559 Chr.2: 185747408-185747408 on Build GRCh38
rs564352476 Chr.2: 185747467-185747467 on Build GRCh38
rs536629449 Chr.2: 185747300-185747300 on Build GRCh38

View Full Product Details