Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66487159-66487426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP3
Entrez Gene ID
10072
Gene Name
dipeptidyl peptidase 3
Gene Aliases
DPPIII
UniGene
Hs.502914
Gene Chromosome Location
Chr.11: 66480013-66509659 on Build GRCh38
Transcripts
NM_001256670.1 NM_005700.4 NM_130443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232203 c.569C>T p.S190L Chr.11: 66487338-66487338 on Build GRCh38
7278676 c.542C>T p.A181V Chr.11: 66487311-66487311 on Build GRCh38
7428417 c.501A>T p.G167G Chr.11: 66487270-66487270 on Build GRCh38
5784201 c.564G>C p.L188L Chr.11: 66487333-66487333 on Build GRCh38
7365397 c.533T>C p.M178T Chr.11: 66487302-66487302 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009198919 Chr.11: 66487407-66487407 on Build GRCh38
rs780614192 Chr.11: 66487262-66487262 on Build GRCh38
rs774235423 Chr.11: 66487340-66487340 on Build GRCh38
rs753149774 Chr.11: 66487366-66487367 on Build GRCh38
rs200764392 Chr.11: 66487379-66487379 on Build GRCh38
rs377309985 Chr.11: 66487373-66487373 on Build GRCh38
rs111809013 Chr.11: 66487279-66487279 on Build GRCh38
rs151083592 Chr.11: 66487327-66487327 on Build GRCh38
rs953000815 Chr.11: 66487208-66487208 on Build GRCh38
rs199659733 Chr.11: 66487364-66487364 on Build GRCh38
rs115287060 Chr.11: 66487187-66487187 on Build GRCh38
rs899999063 Chr.11: 66487221-66487221 on Build GRCh38
rs772587186 Chr.11: 66487361-66487361 on Build GRCh38
rs754200219 Chr.11: 66487245-66487245 on Build GRCh38
rs780205123 Chr.11: 66487324-66487324 on Build GRCh38
rs904914738 Chr.11: 66487179-66487179 on Build GRCh38
rs754288288 Chr.11: 66487384-66487384 on Build GRCh38
rs1047072464 Chr.11: 66487186-66487186 on Build GRCh38
rs749635024 Chr.11: 66487325-66487325 on Build GRCh38
rs979204159 Chr.11: 66487358-66487358 on Build GRCh38
rs768287646 Chr.11: 66487331-66487331 on Build GRCh38
rs185425544 Chr.11: 66487211-66487211 on Build GRCh38
rs1052357850 Chr.11: 66487392-66487392 on Build GRCh38
rs200360094 Chr.11: 66487281-66487281 on Build GRCh38
rs543630858 Chr.11: 66487359-66487359 on Build GRCh38
rs575109777 Chr.11: 66487354-66487354 on Build GRCh38
rs758674085 Chr.11: 66487298-66487298 on Build GRCh38
rs200314618 Chr.11: 66487229-66487229 on Build GRCh38
rs892357454 Chr.11: 66487405-66487405 on Build GRCh38
rs760054906 Chr.11: 66487389-66487389 on Build GRCh38
rs769840950 Chr.11: 66487317-66487317 on Build GRCh38
rs532640618 Chr.11: 66487394-66487394 on Build GRCh38
rs752810487 Chr.11: 66487291-66487291 on Build GRCh38
rs755272628 Chr.11: 66487254-66487254 on Build GRCh38
rs201440123 Chr.11: 66487246-66487246 on Build GRCh38
rs1002303949 Chr.11: 66487185-66487185 on Build GRCh38
rs149825536 Chr.11: 66487302-66487302 on Build GRCh38
rs374576058 Chr.11: 66487207-66487207 on Build GRCh38
rs766934220 Chr.11: 66487244-66487244 on Build GRCh38
rs777681023 Chr.11: 66487301-66487301 on Build GRCh38
rs766897292 Chr.11: 66487235-66487235 on Build GRCh38
rs373458634 Chr.11: 66487241-66487241 on Build GRCh38
rs761152368 Chr.11: 66487242-66487242 on Build GRCh38
rs199771002 Chr.11: 66487273-66487273 on Build GRCh38
rs761551518 Chr.11: 66487219-66487219 on Build GRCh38

View Full Product Details