Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46365098-46365343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
UniGene
Hs.252387
Gene Chromosome Location
Chr.22: 46360834-46537502 on Build GRCh38
Transcripts
NM_014246.1 XM_006724383.3 XM_011530554.2 XM_011530555.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1035155 c.8451C>T p.S2817S Chr.22: 46365334-46365334 on Build GRCh38
5859589 c.8524G>A p.A2842T Chr.22: 46365261-46365261 on Build GRCh38
5043391 c.8451C>A p.S2817R Chr.22: 46365334-46365334 on Build GRCh38
4985579 c.8531G>C p.G2844A Chr.22: 46365254-46365254 on Build GRCh38
5902165 c.8528G>A p.R2843K Chr.22: 46365257-46365257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111632833 Chr.22: 46365165-46365165 on Build GRCh38
rs760526562 Chr.22: 46365298-46365298 on Build GRCh38
rs772687610 Chr.22: 46365180-46365180 on Build GRCh38
rs968671335 Chr.22: 46365319-46365319 on Build GRCh38
rs781379391 Chr.22: 46365323-46365323 on Build GRCh38
rs118156355 Chr.22: 46365324-46365324 on Build GRCh38
rs148280744 Chr.22: 46365310-46365310 on Build GRCh38
rs1040418790 Chr.22: 46365209-46365209 on Build GRCh38
rs202034570 Chr.22: 46365253-46365253 on Build GRCh38
rs565912941 Chr.22: 46365159-46365159 on Build GRCh38
rs1000859944 Chr.22: 46365155-46365155 on Build GRCh38
rs564778741 Chr.22: 46365320-46365320 on Build GRCh38
rs1032620577 Chr.22: 46365172-46365172 on Build GRCh38
rs201897962 Chr.22: 46365301-46365301 on Build GRCh38
rs774171527 Chr.22: 46365288-46365288 on Build GRCh38
rs557943609 Chr.22: 46365177-46365177 on Build GRCh38
rs769346241 Chr.22: 46365181-46365181 on Build GRCh38
rs369554869 Chr.22: 46365300-46365300 on Build GRCh38
rs780558309 Chr.22: 46365251-46365251 on Build GRCh38
rs755424854 Chr.22: 46365321-46365321 on Build GRCh38
rs781500064 Chr.22: 46365209-46365210 on Build GRCh38
rs775433357 Chr.22: 46365295-46365295 on Build GRCh38
rs763659307 Chr.22: 46365224-46365224 on Build GRCh38
rs141057075 Chr.22: 46365249-46365249 on Build GRCh38
rs769981080 Chr.22: 46365284-46365284 on Build GRCh38
rs958131569 Chr.22: 46365176-46365176 on Build GRCh38
rs961077887 Chr.22: 46365121-46365121 on Build GRCh38
rs774118766 Chr.22: 46365199-46365199 on Build GRCh38
rs189299064 Chr.22: 46365158-46365158 on Build GRCh38
rs960958049 Chr.22: 46365142-46365142 on Build GRCh38
rs1007905974 Chr.22: 46365154-46365154 on Build GRCh38
rs578127051 Chr.22: 46365186-46365186 on Build GRCh38
rs144094018 Chr.22: 46365250-46365250 on Build GRCh38
rs536541253 Chr.22: 46365135-46365135 on Build GRCh38
rs780426978 Chr.22: 46365314-46365314 on Build GRCh38
rs778871453 Chr.22: 46365182-46365183 on Build GRCh38
rs902450377 Chr.22: 46365164-46365164 on Build GRCh38
rs560283789 Chr.22: 46365117-46365117 on Build GRCh38
rs199879633 Chr.22: 46365214-46365214 on Build GRCh38
rs756961482 Chr.22: 46365244-46365244 on Build GRCh38
rs767410560 Chr.22: 46365212-46365212 on Build GRCh38
rs752172625 Chr.22: 46365213-46365213 on Build GRCh38
rs1012787331 Chr.22: 46365179-46365179 on Build GRCh38
rs542894795 Chr.22: 46365252-46365252 on Build GRCh38
rs993795976 Chr.22: 46365133-46365133 on Build GRCh38
rs375066055 Chr.22: 46365263-46365263 on Build GRCh38
rs759517218 Chr.22: 46365206-46365206 on Build GRCh38
rs776567447 Chr.22: 46365262-46365262 on Build GRCh38
rs371035318 Chr.22: 46365187-46365187 on Build GRCh38
rs889489153 Chr.22: 46365143-46365143 on Build GRCh38
rs757014797 Chr.22: 46365302-46365302 on Build GRCh38
rs774223704 Chr.22: 46365311-46365311 on Build GRCh38
rs767369340 Chr.22: 46365294-46365294 on Build GRCh38
rs779470956 Chr.22: 46365245-46365245 on Build GRCh38
rs9627424 Chr.22: 46365238-46365238 on Build GRCh38
rs1049402392 Chr.22: 46365140-46365140 on Build GRCh38

View Full Product Details